Skip to Content
Merck
All Photos(1)

Key Documents

TR-1003

Sigma-Aldrich

Polybrene Infection / Transfection Reagent

A highly efficient method of gene transfer into mammalian cells leveraging infection with retroviral vectors.

Synonym(s):

1,5-Dimethyl-1,5-diazaundecamethylene polymethobromide, hexamethrine bromide, retroviral infection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122100
eCl@ss:
32160801
NACRES:
NA.75

Quality Level

form

liquid

manufacturer/tradename

Specialty Media

technique(s)

transfection: suitable

Application

Polybrene is a cationic polymer that can greatly enhance the efficiency of the retroviral or lentiviral infection to the mammalian cells. It acts to neutralize the charge repulsion between virions and the cell surface, thereby increasing infection efficiency. The efficiency of retroviral infection is enhanced significantly, 100 to 1,000 fold in some cells, by including polybrene during the infection. Polybrene with a DMSO shock is also used to mediate DNA transfer into a variety of cell types, such as CHO, chicken embryo fibroblasts, NINH-3T3 and Myeloid cells.


Protocol: Retroviral Infection


Recombinant retroviral stocks are prepared by adding 5mls of growth medium with 5% serum to a near confluent monolayer of transfected retroviral packaging cells in a 100mm plate. After 24 hours the medium is removed and filtered through a 0.45um filter.


Cells to be infected with this recombinant retroviral stock are plated at 500,000 cells per 100mm plate in 10mls of complete medium.


24 hours later, remove the growth medium from the cells. Infect cells with 2mls of the viral supernatant (or a dilution of the virus stock into 2mls) in the presence of 5ug to 10ug of polybrene per ml (final concentration). Incubate cells for 3 to 6 hours at 37°C.


Add 8mls of complete medium. Three days after infection, split the cells 1:5 into selection medium.



Protocol: Transfection


Plate cells at approximately 50% confluence in complete growth medium.


18 to 24 hours post plating, prepare the DNA-Medium-Polybrene solution, immediately before using as follows:


Note: Each component must be added in the proper sequence.

1st: Complete growth medium (2mls for a 60mm plate and 3mls for

a 100mm plate) warmed to 37°C.

2nd: Plasmid DNA, 10ng to 10ug. Gently mix.

3rd: Polybrene to a final concentration of 5ug to 10ug per ml. Gently mix


Remove medium from plate and add DNA-Medium-Polybrene solution to cells. Incubate cells at 37°C for 6 to 20 hours with occasional gentle rocking approximately every 1.5 hours for the first 6 hours.


Remove DNA-Medium-Polybrene solution and gently overlay cells with DMSO shock solution (15% DMSO in 1X HBSS: Specialty Media catalog #S-051-D) 3mls per 60mm dish and 4mls per 100mm plate. Manually rock the dish for 10 seconds to evenly distribute the solution, and then incubate the cells for exactly 4 minutes at 37°C.


Immediately remove the DMSO shock solution and gently rinse the cells twice with complete growth medium, 5mls per wash per 60mm dish, 10mls per wash per 100mm dish


Add complete growth medium to the cells.


For Stable transformants, remove the growth media and split the cells 1:5 into selection medium.


For transient expression, remove the growth medium and add fresh growth medium. Harvest cells and/or medium after 24 to 72 hours.


Reagent is supplied filtered through 0.2μm membranes and hydrated with sterile H2O.

Physical form

10mg polybrene per mL sterile Ultra Pure water.

Storage and Stability

Store at -20°C upto 2 years.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhancement and inhibition of avian sarcoma viruses by polycations and polyanions.
K Toyoshima et al.
Virology, 38(3), 414-426 (1969-07-01)
Nam Ji Sung et al.
Bioscience reports, 40(3) (2020-03-07)
Docosahexaenoic acid (DHA) is an omega-3 fatty acid abundant in fish oils. It is known to have an inhibitory effect on various diseases such as inflammation, diabetes, and cancer. Epithelial-to-mesenchymal transition (EMT) is a process that epithelial cells gain migratory
Margaret M McDaniel et al.
Cell reports, 31(5), 107604-107604 (2020-05-07)
Inflammasome activation leads to pyroptotic cell death, thereby eliminating the replicative niche of virulent pathogens. Although inflammasome-associated cytokines IL-1β and IL-18 have an established role in T cell function, whether inflammasome activation in dendritic cells (DCs) is critical for T cell priming
Marta Llaurado Fernandez et al.
Gynecologic oncology, 157(1), 12-20 (2020-01-20)
Low-grade serous ovarian carcinomas (LGSC) are frequently ER/PR positive, though the mechanisms by which ER/PR regulate prognosis or anti-estrogen treatment efficacy are poorly understood. We studied ER/PR expression in LGSC tumors and cell lines to evaluate patient outcomes and cellular
Arash Nanbakhsh et al.
Methods in molecular biology (Clifton, N.J.), 2097, 107-113 (2019-11-30)
Recent advances in cancer immunotherapy emphasize the need for an efficient method to genetically modify effector lymphocytes to express exogenous "synthetic" genes. NK cells represent 10-20% of total lymphocytes in the peripheral blood of humans and play an essential role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service