Skip to Content
Merck
All Photos(1)

Key Documents

SML1099

Sigma-Aldrich

Roflumilast

≥98% (HPLC), powder, phosphodiesterase 4 PDE4 inhibitor

Synonym(s):

3-(Cyclopropylmethoxy)-N-(3,5-dichloro-4-pyridinyl)-4-(difluoromethoxy)benzamide, 3-Cyclopropylmethoxy-4-difluoromethoxy-N-[3,5-dichloropyrid-4-yl]-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14Cl2F2N2O3
CAS Number:
Molecular Weight:
403.21
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Product Name

Roflumilast, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

ClC1=CN=CC(Cl)=C1NC(C2=CC=C(OC(F)F)C(OCC3CC3)=C2)=O

InChI

1S/C17H14Cl2F2N2O3/c18-11-6-22-7-12(19)15(11)23-16(24)10-3-4-13(26-17(20)21)14(5-10)25-8-9-1-2-9/h3-7,9,17H,1-2,8H2,(H,22,23,24)

InChI key

MNDBXUUTURYVHR-UHFFFAOYSA-N

Gene Information

Application

Roflumilast has been used to evaluate its effect on acute lung inflammation and chronic lung changes in models of cigarette exposure in mice. It is also used to investigate its therapeutic potential to ameliorate primary blast-induced deficits in long-term potentiation (LTP).

Biochem/physiol Actions

Roflumilast is a highly potent, orally active, and selective phosphodiesterase 4 (PDE4) inhibitor with an IC50 of 0.8 nM. Roflumilast has anti-inflammatory properties and is used clinically to treat COPD.
Roflumilast is a highly potent, orally active, and selective phosphodiesterase 4 (PDE4) inhibitor.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roflumilast fully prevents emphysema in mice chronically exposed to cigarette smoke.
Martorana P A, et al.
American Journal of Respiratory and Critical Care Medicine, 172(7), 848-853 (2005)
Peirong Liu et al.
The European journal of neuroscience, 56(4), 4317-4332 (2022-06-30)
Postoperative cognitive dysfunction (POCD) is a prevalent disorder after anaesthesia in the elderly patients. Roflumilast (RF), a phosphodiesterase 4 (PDE-4) inhibitor, could improve cognition with no side effects. Here, we sought to explore the efficacy of RF in the improvement
Amparo Buenestado et al.
PloS one, 8(9), e74640-e74640 (2013-09-26)
Roflumilast is the first phosphodiesterase-4 (PDE4) inhibitor to have been approved for the treatment of COPD. The anti-inflammatory profile of PDE4 inhibitors has not yet been explored in human lung tissues. We investigated the effects of roflumilast and its active
Fernando J Martinez et al.
Lancet (London, England), 385(9971), 857-866 (2015-02-17)
Roflumilast reduces exacerbations in patients with severe chronic obstructive pulmonary disease. Its effect in patients using fixed combinations of inhaled corticosteroids and longacting β2 agonists is unknown. We postulated that roflumilast would reduce exacerbations in patients with severe chronic obstructive
Xiaohua Du et al.
Phytotherapy research : PTR, 33(3), 602-609 (2019-01-15)
The principal active component of isoforskolin (ISOF) is from the plant Coleus forskohlii, native to China, which has attracted much attention for its biological effects. We hypothesize that ISOF and forskolin (FSK) pretreatment attenuates inflammation induced by lipopolysaccharide (LPS) related

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service