Skip to Content
Merck
All Photos(5)

Key Documents

D1756

Sigma-Aldrich

Dexamethasone

≥98% (HPLC), powder, NOS induction inhibitor

Synonym(s):

(11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16α-methylprednisolone, Prednisolone F

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29FO5
CAS Number:
Molecular Weight:
392.46
Beilstein:
2066651
EC Number:
MDL number:
UNSPSC Code:
51422306
PubChem Substance ID:
NACRES:
NA.77

Product Name

Dexamethasone, ≥98% (HPLC), powder

Assay

≥98% (HPLC)

form

powder

color

off-white

mp

262-264 °C (lit.)

solubility

methanol: 25 mg/mL

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

storage temp.

2-8°C

SMILES string

C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO

InChI

1S/C22H29FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,17+,19+,20+,21+,22+/m1/s1

InChI key

UREBDLICKHMUKA-CXSFZGCWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dexamethasone is a potent glucocorticoid with minimal to no mineralocorticoid activity. Dexamethasone exhibits pleiotropic effects on multiple signaling pathways, some of its functions include, inhibition of neutrophil migration and reduction of lymphocyte colony proliferation. Research findings show, dexamethasone might prevent high-altitude pulmonary edema by decreasing pulmonary artery pressure.

Application

Dexamethasone has been used:
  • in culture media to induce differentiation of adipose-derived stem cells into osteogenic tissue
  • as a component of adipocyte differentiation medium
  • for in vitro stimulation of primary normal human bronchial epithelial cells (NHBE) and airway smooth muscle (ASM)

Biochem/physiol Actions

Dexamethasone is known to be effective against relapsed multiple myeloma. It is found to decrease inflammation associated with cerebrospinal fluid and central nervous system (CNS) complications, such as hearing loss.
Glucocorticoid anti-inflammatory agent
Glucocorticoid anti-inflammatory agent. Regulates T cell survival, growth, and differentiation. Inhibits the induction of nitric oxide synthase.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

Dexamethasone is soluble in DMSO (25 mg/ml), chloroform or ethanol.

Reconstitution

Dilute 1 mg/ml stock solution 1:50 with sterile media and store frozen in working aliquots to avoid repeated freeze-thaw.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dexamethasone induction of a heat stress response
Guerrero JA, et al.
Methods in Enzymology (2011)
Increased expression of ADAM33 and ADAM8 with disease progression in asthma
Foley SC, et al.
The Journal of Allergy and Clinical Immunology, 119(4), 863-871 (2007)
Yanan Wu et al.
Polymers, 11(5) (2019-05-11)
Many studies about fabricating organic-inorganic composite materials have been carried out in order to mimic the natural structure of bone. Pearl, which has a special block-and-mortar hierarchical structure, is a superior bone repair material with high osteogenic activity, but it
Dexamethasone in adults with bacterial meningitis
De Gans J and Van de BD
The New England Journal of Medicine, 347(20), 1549-1556 (2002)
Canine adipose-derived-mesenchymal stem cells do not lose stem features after a long-term cryopreservation
Martinello T, et al.
Research in Veterinary Science, 91(1), 18-24 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service