Skip to Content
Merck
All Photos(1)

Key Documents

N3290

Sigma-Aldrich

NKH 477

≥98% (HPLC), powder, adenylyl cyclase activator

Synonym(s):

6-[3-(dimethylamino)propionyl]-forskolin, Colforsin dapropate hydrochloride, N,N-dimethyl-, (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-5-(acetyloxy)-3-ethenyldodecahydro-10,10b-dihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-1H-naphtho[2,1-b]pyran-6-yl ester b-Alanine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H43NO8·HCl
CAS Number:
Molecular Weight:
546.09
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

NKH 477, ≥98% (HPLC), powder

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 3 mg/mL, clear to slightly hazy (warmed)

storage temp.

2-8°C

SMILES string

Cl.CN(C)CCC(=O)O[C@@H]1[C@H](OC(C)=O)[C@@]2(C)O[C@](C)(CC(=O)[C@]2(O)[C@@]3(C)[C@@H](O)CCC(C)(C)[C@H]13)C=C

InChI

1S/C27H43NO8.ClH/c1-10-24(5)15-18(31)27(33)25(6)17(30)11-13-23(3,4)21(25)20(35-19(32)12-14-28(8)9)22(34-16(2)29)26(27,7)36-24;/h10,17,20-22,30,33H,1,11-15H2,2-9H3;1H/t17-,20-,21-,22-,24-,25-,26+,27-;/m0./s1

InChI key

VIRRLEDAYYYTOD-YHEOSNBFSA-N

Biochem/physiol Actions

NKH 477 is a potent, orally active adenylyl cyclase activator. It is a water soluble derivative of forskolin, and dose dependentyl increases cAMP and blocks potassium-induced contraction concetrations in smooth muscle strips (IC50 = 80 nM). NKH 477 causes relaxation of histamine treated guinea pig smooth muscle (IC50 = 32 nM).

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Preparation Note

A clear to slightly hazy solution is possible.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Li-Hua Hang et al.
Pharmacological reports : PR, 65(3), 710-716 (2013-08-21)
It has been shown that spinal PKA/CREB signaling pathway is involved in neuropathic and inflammatory pain, but its effects on bone cancer pain have not previously been investigated. The aim of this study was to examine the potential role of
Saw Marlar et al.
American journal of physiology. Cell physiology, 306(6), C598-C606 (2014-01-24)
Regulated urine concentration takes place in the renal collecting duct upon arginine vasopressin (AVP) stimulation, where subapical vesicles containing aquaporin-2 (AQP2) are inserted into the apical membrane instantly increasing water reabsorption and urine concentration. The reabsorped water exits via basolateral
Mika Shirasu et al.
Neuron, 81(1), 165-178 (2013-12-24)
Musk odorants are used widely in cosmetic industries because of their fascinating animalic scent. However, how this aroma is perceived in the mammalian olfactory system remains a great mystery. Here, we show that muscone, one musk odor secreted by various
Chin-San Loo et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(42), 17119-17124 (2013-10-03)
In kidney collecting duct cells, filamentous actin (F-actin) depolymerization is a critical step in vasopressin-induced trafficking of aquaporin-2 to the apical plasma membrane. However, the molecular components of this response are largely unknown. Using stable isotope-based quantitative protein mass spectrometry
Mika Takarada-Iemata et al.
Journal of neurochemistry, 130(3), 374-387 (2014-04-05)
N-myc downstream-regulated gene 2 (Ndrg2) is a differentiation- and stress-associated molecule predominantly expressed in astrocytes in the CNS. In this study, we examined the expression and the role of Ndrg2 after cortical stab injury. We observed that Ndrg2 expression was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service