Skip to Content
Merck

Skip To

A265

ARL 67156 trisodium salt hydrate

≥98% (HPLC), solid, ecto-ATPase inhibitor

Synonym(s):

6-N,N-Diethyl-β-γ-dibromomethylene-D-adenosine-5′-triphosphate trisodium salt hydrate, FPL 67156

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C15H21Br2N5Na3O12P3 · xH2O
CAS Number:
Molecular Weight:
785.05 (anhydrous basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

ARL 67156 trisodium salt hydrate, ≥98% (HPLC), solid

Quality Segment

assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

H2O: >20 mg/mL, DMSO: insoluble

storage temp.

−20°C

SMILES string

O.[Na+].[Na+].[Na+].CCN(CC)c1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)C(Br)(Br)P([O-])([O-])=O)[C@@H](O)[C@H]3O

InChI

1S/C15H24Br2N5O12P3.3Na.H2O/c1-3-21(4-2)12-9-13(19-6-18-12)22(7-20-9)14-11(24)10(23)8(33-14)5-32-37(30,31)34-36(28,29)15(16,17)35(25,26)27;;;;/h6-8,10-11,14,23-24H,3-5H2,1-2H3,(H,28,29)(H,30,31)(H2,25,26,27);;;;1H2/q;3*+1;/p-3/t8-,10-,11-,14-;;;;/m1..../s1

InChI key

UWMONIJVKGTUGE-OPKBHZIBSA-K

Gene Information

human ... ENTPD2(954)

General description

ARL 67156, a nucleotide/ATP analog is a competitive inhibitor of CD39. This ecto-ATPase inhibitor is presumed to be metabolically stable in biological studies due to its β, γ-dibromomethylene bridge.
ARL-67156 preferentially inhibits the degradation of ADP compared to the degradation of ATP in the murine colon. It is commonly employed to inhibit ATP hydrolysis in tissue preparations.

Application

ARL 67156 trisodium salt hydrate has been used:
  • to prevent ATP degradation during cerebrospinal fluid extraction
  • as an ATPase inhibitor in ATP quantification assay for preventing ATP degradation in cytoplasm and mitochondrion
  • as an ecto-ATPase inhibitor to treat cancer cells for ATP release assay

Biochem/physiol Actions

ecto-ATPase inhibitor; prevents metabolism of P2 purinoceptor agonists.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A166A2846M3808
description

≥98% (HPLC), solid

description

≥98% (HPLC), solid

description

≥98% (HPLC), solid

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

solid

form

solid

form

solid

form

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

storage temp.

−20°C

solubility

H2O: >20 mg/mL, DMSO: insoluble

solubility

H2O: soluble

solubility

H2O: soluble

solubility

DMSO: 1.1 mg/mL, H2O: >10 mg/mL

color

white to off-white

color

white

color

off-white to light tan

color

-


Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What is the stability of Product A265, ARL 67156 trisodium salt hydrate, in solution (water)?

    1 answer
    1. The stability of this product in solution has not been determined. Various sources suggest that stock solutions may be stored in aliquots at ?20°C for 1 month and at ?80°C for 6 months. This information has not been validated.

      Helpful?

Reviews

No rating value

Active Filters