Skip to Content
Merck
All Photos(2)

Documents

SMB00610

Sigma-Aldrich

Lipopolysaccharide from Porphyromonas gingivalis

purified by phenol extraction

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

biological source

Porphyromonas gingivalis

Quality Level

form

powder

purified by

phenol extraction

impurities

≤3% Protein (Lowry)

color

white to faint yellow

solubility

triethylene glycol dimethyl ether: 0.90-1.10 mg/mL to hazy, colorless to light yellow

shipped in

ambient

storage temp.

2-8°C

General description

Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria like Porphyromonas gingivalis. It differs from LPS from Escherichia coli in its structure and various functional activities.

Application

Lipopolysaccharide has been used:

  • as a treatment to stimulate acute lung injury in human bronchial epithelial (HBE) cells
  • as an agonist for toll-like receptor 4 (TLR4) and to stimulate intestinal normal fibroblasts (NFs) to study osteopontin (OPN) expression in myofibroblasts
  • to study its effects on pro-inflammatory and pro-coagulant genes expression in endothelial cells

Biochem/physiol Actions

LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular patterns (PAMPs).

Additionally, it was demonstrated that the mechanisms by which LPS from E. coli and P. gingivalis modulate cluster of differentiation 14 (CD14), toll-like receptor 2 (TLR2), and toll-like receptor 4 (TLR4) surface expression, primary and secondary cytokine responses are different.

Porphyromonas gingivalis is a Gram-negative bacterium that is known to be involved in adult periodontitis. Periodontitis is a chronic inflammatory disease characterized by the recession of the supportive tissue surrounding teeth. Studies have shown that the LPS from P. gingivalis plays an important role in this disease.
A recent study demonstrated that LPS from P. gingivalis stimulates insulin secretion by the pancreatic β cell line, MIN6. In the presence of 5 mM glucose and 50-500 ng/mL LPS from P. gingivalis, a significant induction of insulin secretion was observed.

Preparation Note

LPS from P. gingivalis is soluble in water and cell culture medium DMEM (1-5 mg/mL), yielding a clear solution.

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differential expression of immunoregulatory genes in monocytes in response to Porphyromonas gingivalis and Escherichia coli lipopolysaccharide.
Barksby, H.E, et al.
Clinical and Experimental Immunology, 156, 479-487 (2009)
Virulence factors of Porphyromonas gingivalis.
Holt, S.C., et al.
Periodontology 2000, 20, 168-238 (1999)
Diya Zhang et al.
Innate immunity, 14(2), 99-107 (2008-08-21)
Lipopolysaccharide (LPS) derived from the periodontal pathogen Porphyromonas gingivalis has been shown to differ from enterobacterial LPS in structure and function; therefore, the Toll-like receptors (TLRs) and the intracellular inflammatory signaling pathways are accordingly different. To elucidate the signal transduction
Epidemiology of subgingival bacterial pathogens in periodontal disease
Zambon, J. J., et al.
Molecular Pathogenesis of Periodontal Disease, 1, 1-12 (1994)
Epidemiology of subgingival bacterial pathogens in periodontal disease.
Zambon, J.J., et al.
Molecular Pathogenesis of Periodontal Disease, 1-12 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service