Skip to Content
Merck
All Photos(1)

Documents

L7770

Sigma-Aldrich

Lipopolysaccharides from Salmonella enterica serotype enteritidis

γ-irradiated, BioXtra, suitable for cell culture

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201
NACRES:
NA.75

biological source

Salmonella enterica (Serotype enteritidis)

Quality Level

sterility

γ-irradiated

product line

BioXtra

form

lyophilized powder

purified by

gel-filtration chromatography

technique(s)

cell culture | mammalian: suitable

impurities

<1% Protein (Lowry)

color

white to yellow cast

solubility

water: 4.90-5.10 mg/mL, faintly hazy to hazy, colorless

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Lipopolysaccharides from Salmonella enterica serotype enteritidis are used in cell culture and cell binding applications. It was used to study the role of inflammation in the progression of neuropathic pain and aneurysm.
Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4).

Biochem/physiol Actions

Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs).

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosemary Fernandez et al.
Bio-protocol, 5(11) (2016-05-03)
The ability to assess the function of a range of cytokine, antigen receptor, and Toll-like receptor (TLR) signaling pathways in a range of immune cells could provide a kind of fingerprint of the state of the human immune system. The
Elisabetta Liverani et al.
Journal of leukocyte biology, 95(2), 313-323 (2013-10-22)
Thienopyridines are a class of antiplatelet drugs that are metabolized in the liver to several metabolites, of which only one active metabolite can irreversibly antagonize the platelet P2Y12 receptor. Possible effects of these drugs and the role of activated platelets
Anita Mikołajczyk et al.
Gut pathogens, 9, 73-73 (2017-12-20)
The present research was conducted to investigate the influence of a low, single dose of LPS, which does not result in any clinical symptoms of intoxication on the expression of selected neuropeptides within the intestines of the domestic pig. This
Christopher J Neufeldt et al.
PLoS pathogens, 9(10), e1003744-e1003744 (2013-11-10)
Hepatitis C virus (HCV) infection induces formation of a membranous web structure in the host cell cytoplasm where the viral genome replicates and virions assemble. The membranous web is thought to concentrate viral components and hide viral RNA from pattern
Rosemary Fernandez et al.
Bio-protocol, 5(11) (2016-07-23)
Phosphorylation of tyrosine, serine, and threonine residues is critical for the control of protein activity involved in various cellular events. An assortment of kinases and phosphatases regulate intracellular protein phosphorylation in many different cell signaling pathways. These pathways include T

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service