Skip to Content
Merck
All Photos(3)

Documents

L2143

Sigma-Aldrich

Lipopolysaccharides from Escherichia coli K-235

purified by phenol extraction

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

Escherichia coli (K-235)

Quality Level

form

lyophilized powder

purified by

phenol extraction

impurities

<3% Protein (Lowry)

color

white to faint yellow

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is phenol extracted from E. coli serotype K235. The source strain is ATCC 13027.

Application


  • Establishment of disseminated intravascular coagulation (DIC) model by a single iv administration of Escherichia coli-derived lipopolysaccharide (LPS) to cynomolgus monkeys and evaluation of its pathophysiological status.: This study established a DIC model in cynomolgus monkeys using E. coli-derived LPS, providing insights into the pathophysiological status and potential therapeutic interventions for DIC (Minomo et al., 2017).

  • Different bacterial lipopolysaccharides as toxicants and stressors in the shrimp Palaemon elegans.: This research explored the toxic effects of various bacterial LPS, including E. coli K-235, on the shrimp Palaemon elegans, advancing the understanding of LPS-induced stress responses in marine organisms (Lorenzon et al., 2002).

Biochem/physiol Actions

Lipopolysaccharides (LPS) are localized in the outer layer of the membrane and are, in noncapsulated strains, exposed on the cell surface. They contribute to the integrity of the outer membrane, and protect the cell against the action of bile salts and lipophilic antibiotics.

Preparation Note

The product is soluble in water (5 mg/ml) or cell culture medium (1 mg/ml) yielding a hazy, faint yellow solution. A more concentrated, though still hazy, solution (20 mg/ml) has been achieved in aqueous saline after vortexing and warming to 70-80 oC. Lipopolysaccharides are molecules that form micelles in every solvent. Hazy solutions are observed in water and phosphate buffered saline. Organic solvents do not give clearer solutions. Methanol yields a turbid suspension with floaters, while water yields a homogeneously hazy solution.

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jay W Chaplin et al.
The Journal of experimental medicine, 210(10), 2135-2146 (2013-09-11)
Antigen (Ag) targeting is an efficient way to induce immune responses. Ag is usually coupled to an antibody (Ab) specific for a receptor expressed on dendritic cells (DCs), and then the Ag-anti-receptor is inoculated with an adjuvant. Here we report
Agnieszka Zdybicka-Barabas et al.
Amino acids, 51(2), 175-191 (2018-09-01)
Cecropins constitute an important family of insect antimicrobial peptides involved in humoral innate immune response. In comparison with the highly basic cecropins A and B, cecropins D are less cationic and more hydrophobic. Interestingly, cecropins D were described only in
Jessica F Hoffman et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(39), 10039-10049 (2016-09-30)
Identifying and understanding critical periods in brain development is essential to decoding the long-term impact of widespread, poorly defined, and frequently occurring insults such as inflammation. Using the laboratory rat Rattus norvegicus, we have discovered a narrowly constrained critical period
Stefka Gyoneva et al.
Glia, 62(8), 1345-1360 (2014-05-09)
Microglia, the resident immune cells of the central nervous system, exist in either a "resting" state associated with physiological tissue surveillance or an "activated" state in neuroinflammation. We recently showed that ATP is the primary chemoattractor to tissue damage in
Long-Yun Zhou et al.
Journal of neurochemistry, 155(2), 154-176 (2020-03-28)
Cervical spondylotic myelopathy (CSM) is a common cause of disability with few treatments. Aberrant mitochondrial dynamics play a crucial role in the pathogenesis of various neurodegenerative diseases. Thus, regulation of mitochondrial dynamics may offer therapeutic benefit for the treatment of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service