Skip to Content
Merck
All Photos(1)

Key Documents

L8643

Sigma-Aldrich

Lipopolysaccharides from Pseudomonas aeruginosa 10

purified by gel-filtration chromatography

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

Pseudomonas aeruginosa (10)

Quality Level

form

lyophilized powder

purified by

gel-filtration chromatography

impurities

<3% Protein (Lowry)

color

white to faint yellow

solubility

water: 4.90-5.10 mg/mL, cloudy to hazy, colorless to faintly yellow

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs).
Lipopolysaccharides from Pseudomonas aeruginosa 10 has been used as an inflammatory mediator in cells.

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clara cells attenuate the inflammatory response through regulation of macrophage behavior.
Snyder JC
American Journal of Respiratory Cell and Molecular Biology, 42, 161-171 (2010)
Macrophages directly contribute to the exaggerated inflammatory response in cystic fibrosis transmembrane conductance regulator-/- mice.
Bruscia EM
American Journal of Respiratory Cell and Molecular Biology, 40, 295-295 (2009)
Sandra Chang et al.
PloS one, 10(7), e0131810-e0131810 (2015-07-07)
Oxidant production from DUOX1 has been proposed to lead to neutrophil recruitment into the airways when lung homeostasis is compromised. The objective of this study was to determine whether DUOX-derived hydrogen peroxide is required for LPS-induced neutrophil recruitment, using a
Coordinate induction of two antibiotic genes in tracheal epithelial cells exposed to the inflammatory mediators lipopolysaccharide and tumor necrosis factor alpha.
Russell JP
Infection and Immunity, 64, 1565-1565 (1996)
Austin Boucher et al.
Journal of immunology (Baltimore, Md. : 1950), 206(3), 540-553 (2020-12-18)
Macrophages are critical for regulating inflammatory responses. Environmental signals polarize macrophages to either a proinflammatory (M1) state or an anti-inflammatory (M2) state. We observed that the microRNA (miRNA) cluster mirn23a, coding for miRs-23a, -27a, and -24-2, regulates mouse macrophage polarization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service