Skip to Content
Merck
All Photos(1)

Documents

L5293

Sigma-Aldrich

Lipopolysaccharides from Escherichia coli O111:B4

Ready Made solution, 1 mg/mL

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

Escherichia coli (O111:B4)

Quality Level

form

aqueous solution

concentration

1 mg/mL

color

colorless to faint yellow

solubility

water: soluble

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Lipopolysaccharides from Escherichia coli O111:B4 has been used:
  • to stimulate an inflammatory response in porcine liver tissues in-vivo and piglet ileum tissues ex vivo
  • to stimulate Ex vivo natural killer(NK) cells and hepatic B cells
  • to induce arthritis in a standard rabbit model

Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs).

Biochem/physiol Actions

Lipopolysaccharides (LPS) are distributed on the outer layer of the membrane and in non-capsulated strains, present on the cell surface. LPS maintains the outer membrane integrity. In addition, it protects the cell from bile salts stress and lipophilic antibiotics.

Preparation Note

The product is soluble in water (5 mg/ml) or cell culture medium (1 mg/ml) yielding a hazy, faint yellow solution. A more concentrated, though still hazy, solution (20 mg/ml) has been achieved in aqueous saline after vortexing and warming to 70-80 oC. Lipopolysaccharides are molecules that form micelles in every solvent. Hazy solutions are observed in water and phosphate buffered saline. Organic solvents do not give clearer solutions. Methanol yields a turbid suspension with floaters, while water yields a homogeneously hazy solution.

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NKG2D is critical for NK cell activation in host defense against Pseudomonas aeruginosa respiratory infection
Wesselkamper S, et al.
Journal of immunology (Baltimore, Md. : 1950), 181(8), 5481-5489 (2008)
Effects of dietary beta-glucans supplementation on cytokine expression in porcine liver
Ryan M, et al.
Journal of Animal Science, 90(suppl-4), 40-42 (2012)
Effect of maternal seaweed extract supplementation on suckling piglet growth, humoral immunity, selected microflora, and immune response after an ex vivo lipopolysaccharide challenge
Leonard SG, et al.
Journal of Animal Science, 90(2), 505-514 (2012)
6 Analysis of Lipopolysaccharides of Gram-Negative Bacteria
Methods in Microbiology, 18(5), 157-207 (1985)
Imaging of normal and pathologic joint synovium using nonlinear optical microscopy as a potential diagnostic tool
Tiwari N, et al.
Journal of Leukocyte Biology, 15(5), 056001-056001 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service