Skip to Content
Merck
All Photos(2)

Key Documents

S5193

Sigma-Aldrich

SYBR® Green JumpStart Taq ReadyMix

for quantitative PCR, without MgCl2 in buffer

Synonym(s):

Hot start PCR master mix, sybr green PCR master mix, sybr green qPCR

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106300
NACRES:
NA.55

form

liquid

usage

sufficient for 100 reactions
sufficient for 20 reactions
sufficient for 400 reactions

feature

dNTPs included
hotstart

concentration

1.25 units/reaction (50 μL reaction volume)

technique(s)

qPCR: suitable

color

colorless

input

purified DNA

suitability

suitable for PCR

application(s)

agriculture

compatibility

Bio-Rad MyiQ
Bio-Rad iCycler iQ
Bio-Rad iQ 5
for use with ABI 5700
for use with ABI 7000
for use with ABI 7300
for use with ABI 7500 Fast
for use with ABI 7500
for use with ABI 7700
for use with ABI 7900 Fast
for use with ABI 7900 HT
for use with ABI 7900
for use with ABI StepOne
for use with ABI StepOnePlus
for use with ABI ViiA 7
for use with Bio-Rad CFX384
for use with Bio-Rad CFX96
for use with Bio-Rad MJ Chromo4
for use with Bio-Rad MJ Opticon 2
for use with Bio-Rad MJ Opticon Cepheid SmartCycler
for use with Bio-Rad MJ Opticon
for use with Bio-Rad MiniOpticon
for use with Eppendorf® Mastercycler ep realplex2 s
for use with Eppendorf® Mastercycler ep realplex
for use with Illumina Eco qPCR
for use with Qiagen Corbett Rotor-Gene 3000
for use with Qiagen Corbett Rotor-Gene 6000
for use with Qiagen Corbett Rotor-Gene Q
for use with Roche LightCycler 480
for use with Strategene Mx3000P
for use with Strategene Mx3005P
for use with Strategene Mx4000

detection method

SYBR® Green

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

SYBR® Green JumpStart Taq ReadyMix combines the performance enhancements of JumpStart Taq antibody for hot start PCR with SYBR Green I and the convenience of an easy-to-use ReadyMix solution. This ready-to-use mixture of SYBR Green I, JumpStart Taq DNA polymerase, 99% pure deoxynucleotides and reaction buffer is provided in a 2× concentrate for ease of use. Simply add 25 μL of the 2× mix to DNA template, primers and water. The JumpStart Taq antibody inactivates DNA polymerase at room temperature. When the temperature is raised above 70 °C in the first denaturation step of the cycling process, the complex dissociates and the polymerase becomes fully active. JumpStart Taq DNA polymerase prevents non-specific amplification resulting in more accurate CT values.

Application

SYBR® Green JumpStart Taq ReadyMix has been used:

  • During RNA extraction, reverse transcription and Quantitative PCR
  • During analysis of leptin and leptin receptor expression. (PCR and melting curve analyses were performed)
  • For tissue RNA Isolation. (PCR and melting curve analyses were performed)
  • in real-time PCR (RT-PCR) of reverse-transcribed cDNA template
  • For qPCR amplifications using SYBR
  • For multiplex PCR
SYBR® Green JumpStart Taq ReadyMix has been used:
  • to prepare the real-time PCR mixture
  • in the quantitative analysis of gene expression by real time, quantitative polymerase chain reaction (qPCR)
  • to determine the mRNA expression of nuclear factor erythroid 2 (NFE2)-related factor 2 (Nrf2) gene by qPCR

Features and Benefits

  • Delivers the benefits of antibody-inactivated hot-start PCR with SYBR Green detection in a ReadyMix ideal for high throughput applications; only primers and template are required
  • JumpStart Taq DNA polymerase prevents amplification of non-specific products, resulting in increased efficiency and higher target yield
  • SYBR® Green JumpStart Taq ReadyMix for SYBR based qPCR is formulated with MgCl2 or packaged with a separate vial for ease of optimization. ReadyMixes are compatible with tube- and plate-based instruments
  • This master mix allows consistency from one reaction to the next
  • Designed to minimize contamination
  • Reduced primer dimers
  • Reduced set-up time as compared to manual or wax HotStart® methods
  • Allows for room temperature set-up and contains a fluorescent dye, which is ideal for qPCR applications

Packaging

Default reaction volume is 50 μL

20RXN is packaged as 1 X 500 μL
100RXN is packaged as 1 X 2.5 mL
400RXN is packaged as 1 X 10 mL
500RXN is packaged as 1 X 12.5 mL

Other Notes

Magnesium chloride is provided separately for optimization.

Principle

SYBR Green JumpStart Taq ReadyMix is recommended for single product real-time amplification experiments and can also be used for PCR optimization prior to manufacture of fluorescent-labeled probes. Fluorescent labeled probes are not recommended for use with SYBR Green I dye.

SYBR Green I, a commonly used fluorescent DNA binding dye, binds all double-stranded DNA and detection is monitored by measuring the increase in fluorescence throughout the cycle. SYBR Green I has an excitation and emission maxima of 494 nm and 521 nm, respectively. The instrument settings for ROX reference dye are satisfactory for the measurement of the Reference Dye for Quantitative PCR. Specificity of Sigma′s SYBR based QPCR detection is greatly enhanced by the incorporation of a hot-start mediated taq polymerase, JumpStart Taq.

Legal Information

Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,994,056 and 6,171,785.. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim (such as apparatus or system claims in US Patent No. 6,814,934) and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Eppendorf is a registered trademark of Eppendorf AG
HOTSTART is a registered trademark of Molecular BioProducts, Inc.
JumpStart is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC
SYBR is a registered trademark of Life Technologies

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nesrine S El Sayed et al.
Frontiers in pharmacology, 12, 666502-666502 (2021-08-10)
Sporadic Alzheimer's disease (AD) is the most common neurodegenerative disorder with cognitive dysfunction. Remarkably, alteration in the gut microbiome and resultant insulin resistance has been shown to be connected to metabolic syndrome, the crucial risk factor for AD, and also
Brown fat like gene expression in the epicardial fat depot correlates with circulating HDL-cholesterol and triglycerides in patients with coronary artery disease
Chechi K, et al.
International Journal of Cardiology, 167(5), 2264-2270 (2013)
Nesrine S El Sayed et al.
Oxidative medicine and cellular longevity, 2021, 5525306-5525306 (2021-07-27)
Alzheimer's disease (AD) is a neurodegenerative disease characterized by cognitive impairment. Gut microbiota dysfunction (dysbiosis) is implicated in the pathology of AD and is associated with several detrimental consequences, including neurotransmitter depletion, oxidative stress, inflammation, apoptosis, and insulin resistance, which
Wael Fathy et al.
Journal of basic microbiology, 61(4), 330-338 (2021-02-19)
Microalgae have received continued attention as a potential source for biofuel production. However, the lack of suitable strains that provide a lipid-rich biomass and tolerate harsh condition inhibits their industrial application. This report describes an effort to transform Synechocystis sp.
Wael Fathy et al.
Journal of basic microbiology, 61(4), 330-338 (2021-02-19)
Microalgae have received continued attention as a potential source for biofuel production. However, the lack of suitable strains that provide a lipid-rich biomass and tolerate harsh condition inhibits their industrial application. This report describes an effort to transform Synechocystis sp.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service