Skip to Content
Merck
All Photos(2)

Key Documents

S9430

Sigma-Aldrich

SYBR® Green I nucleic acid gel stain

greener alternative

10,000 × in DMSO

Synonym(s):

DNA stain, SYBR® green gel dye, safer gel stain

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.52
Pricing and availability is not currently available.

form

solution

Quality Level

usage

1.0 mL sufficient for 100 mini-gels

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

10,000 × in DMSO

technique(s)

PCR: suitable

greener alternative category

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

SYBR® Green I is a proprietary asymmetrical cyanine dye, which is used to detect nucleic acids. It consists of a N-alkylated benzothiazolium or benzoxazolium ring system, that is joined by a monomethine bridge to a pyridinium or quinolinium ring system.[1] SYBR Green I binds to the minor groove of dsDNA and is excited at a wavelength of 480 nm. It has a peak fluorescence emission of 520 nm.[2]

Application

SYBR® Green I nucleic acid gel stain has been used for:
  • the quantification of dsDNA[3]
  • to stain DNA in polymerase chain reaction (PCR)[4]
  • for comet assay technique[5]
  • to assess spermatozoon membrane integrity[6]
  • for visual inspection of DNA amplified by loop-mediated isothermal amplification (LAMP)[7]
  • as a fluorescent dye in flow cytometry[8]
  • real-time quantitative reverse transcription-polymerase chain reaction (qRT-PCR) for staining reverse-transcribed cDNA[8]

Features and Benefits

  • An ultrasensitive stain for post-electrophoresis staining of dsDNA in agarose or polyacrylamide gels
  • It can also detect ssDNA and RNA in denaturing agarose/formaldehyde and polyacrylamide/urea gels without any pre-washing steps
  • It is less mutagenic than ethidium bromide in Ames tests
  • It provides 50-100 times greater detection sensitivity than ethidium bromide for oligonucleotides
  • Useful for many applications with a limited amount of DNA
  • The binding of SYBR® Green I to DNA does not inhibit the activity of many common restriction endonucleases, including Hind III and EcoR I
  • Removal of this stain in-gel digestion and ligation techniques is not needed
  • SYBR Green I is a greener alternative product to ethidium bromide for staining

Storage and Stability

Store the product at –20 °C. The diluted Staining Solution may be stored, protected from light, either at 2–8 °C for several weeks or at room temperature for 3–4 days.

Legal Information

SYBR is a registered trademark of Life Technologies

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advances in Clinical Chemistry (2005)
Model of vitamin and mineral deficiency for toxicological research: Apoptosis activity under conditions of CCL4 intoxication
Tyshko NV and Shestakova SI
Toxicology Reports, 6, 151-154 (2019)
Cristina Molnar et al.
Science advances, 5(8), eaaw7965-eaaw7965 (2019-08-28)
The notable male predominance across many human cancer types remains unexplained. Here, we show that Drosophila l(3)mbt brain tumors are more invasive and develop as malignant neoplasms more often in males than in females. By quantitative proteomics, we have identified
Multiplex Polymerase Chain Reaction (PCR) and Real-Time
Multiplex PCR for the Simultaneous Detection of Plant Viruses
V. Pallas
Methods in Molecular Biology (2009)
Comparison between Flow Cytometry, Microscopy, and Lactate Dehydrogenase-Based Enzyme-Linked Immunosorbent Assay for Plasmodium falciparum Drug Susceptibility Testing under Field Conditions
Charles J. Woodrow
Journal of Clinical Microbiology (2015)

Questions

1–4 of 4 Questions  
  1. Does Millipore Sigma offer SAFE Red loading dye?

    1 answer
    1. SAFE Red loading dye is not offered by Millipore Sigma.

      Helpful?

  2. What is the molarity of this product?

    1 answer
    1. This product is a 10,000X concentrated solution suitable to prepare 10L of staining solution for in-gel staining of DNA and RNA. The specific concentration is considered proprietary. Published working concentrations for solutions of Sybr Green I range from 0.5 - 1.0 ug/ml. The molecular weight as published by Chebi, is 509.74 g/mol. Please see the link below to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/282/803/s9430pis-ms.pdf

      Please see the link below to review the Chebi profile:
      https://www.ebi.ac.uk/chebi/searchId.do?chebiId=CHEBI:51461


      chebi file
      https://www.ebi.ac.uk/chebi/searchId.do?chebiId=CHEBI:51461 MW 509.74
      Science direct
      https://www.sciencedirect.com/topics/biochemistry-genetics-and-molecular-biology/sybr-green-i
      'working' concentration 0.5 - 1.0 ug/ml

      Helpful?

  3. Will Product S9430, SYBR® Green I nucleic acid gel stain, stain DNA that contains deaza-G modified nucleotides in place of guanisine?

    1 answer
    1. Product S9430, SYBR® Green I nucleic acid gel stain, will poorly stain DNA containing deaza-G modified nucleotides in place of guanisine.  A way around this issue would be to use a mixture of deaza-G modified nucleotides with guanisine.  This will allow for better staining.

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service