Skip to Content
Merck
All Photos(3)

Key Documents

KCQS02

Sigma-Aldrich

KiCqStart® SYBR® Green qPCR ReadyMix

with ROX for ABI instruments

Synonym(s):

qPCR master mix, sybr green qPCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

form

liquid

usage

sufficient for 1250 reactions
sufficient for 250 reactions
sufficient for 5000 reactions

feature

dNTPs included
hotstart

storage condition

protect from light

technique(s)

qPCR: suitable

color

colorless

input

purified DNA

compatibility

for use with ABI 5700
for use with ABI 7000
for use with ABI 7300
for use with ABI 7700
for use with ABI 7900 HT Fast
for use with ABI 7900 HT
for use with ABI 7900
for use with ABI StepOne
for use with ABI StepOnePlus

detection method

SYBR® Green

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

KiCqStart SYBR Green qPCR ReadyMix is a 2X concentrated, ready-to-use master mix that contains all components, except primers and template, for real-time quantitative PCR (qPCR) This unique combination of proprietary buffer, stabilizers, and Hot-Start Taq DNA polymerase delivers maximum PCR efficiency, sensitivity, specificity and robust fluorescent signal using fast, or conventional, cycling protocols with SYBR Green qPCR.

Highly specific amplification is crucial to successful qPCR with SYBR Green I dye technology because this dye binds to and detects any dsDNA generated during amplification. Hot-Start Taq DNA polymerase is antibody mediated to be inactive prior to the initial PCR denaturation step.

Application

KiCqStart® SYBR® Green qPCR ReadyMix has been used:

  • in the amplification and quantification of cDNA reverse transcribed from RNA extracted from mice brain samples in a 2-step RT-qPCR assay
  • to analyze DNA purified by ChIP technique
  • to perform gene expression analysis
  • in amplification of RNA isolated from hearts of adult TL wild-type fish by quantitative real-time polymerase chain reaction (PCR)
PCR applications:
  • Gene expression
  • DNA quantification
  • CHiP

Features and Benefits

  • Assay results in as little as 33 minutes
  • Highly efficient and sensitive real-time PCR results
  • Little/no optimization required

Components

2X reaction buffer containing optimized concentrations of MgCl2, dNTPs, (dATP, dCTP, dGTP, dTTP), KiCqStart Taq DNA Polymerase, SYBR Green dye, ROX Reference Dye (for 580-585 nm excitation), and stabilizers.

packaging:
250 reactions* = 2 X 1.25 mL tubes
1250 reactions* = 10 X 1.25 mL tubes
5000 reactions* = 1 X 50 mL tube
*number of reactions based on a 20uL volume

Other Notes

Storage Conditions:
KiCqStart SYBR Green qPCR ReadyMix is stable for 1 year when stored in a constant temperature freezer at -20°C, protected from light. For convenience, it may be stored unfrozen at +2 to +8°C for up to 6 months. After thawing, mix thoroughly before using. Repeated freezing and thawing of the product is not recommended. However, the product demonstrated no loss of performance after 20 freeze-thaw cycles or 2 months at +20°C.

Legal Information

KiCqStart is a registered trademark of QIAGEN Beverly Inc.
ROX is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
ReadyMix is a trademark of Sigma-Aldrich Co. LLC
SYBR is a registered trademark of Life Technologies

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gene expression study of phase I and II metabolizing enzymes in RPTEC/TERT1 cell line: application in in vitro nephrotoxicity prediction.
Shah H
Xenobiotica, 3, 1-7 (2016)
MiR-219a-5p Enriched Extracellular Vesicles Induce OPC Differentiation and EAE Improvement More Efficiently Than Liposomes and Polymeric Nanoparticles
<BIG><BIG>Osorio QI, et al.</BIG></BIG>
Pharmaceutics, 12, 186-186 (2020)
Gene expression study of phase I and II metabolizing enzymes in RPTEC/TERT1 cell line: application in in vitro nephrotoxicity prediction
<BIG><BIG>Shah H, et al.</BIG></BIG>
Xenobiotica, 47, 837-843 (2017)
Absence of the neurogenesis-dependent nuclear receptor TLX induces inflammation in the hippocampus
<BIG><BIG>Kozareva DA, et al.</BIG></BIG>
Journal of Neuroimmunology, 331, 87-96 (2019)
S M Kamel et al.
Nature communications, 12(1), 7151-7151 (2021-12-11)
The heterozygous Phospholamban p.Arg14del mutation is found in patients with dilated or arrhythmogenic cardiomyopathy. This mutation triggers cardiac contractile dysfunction and arrhythmogenesis by affecting intracellular Ca2+ dynamics. Little is known about the physiological processes preceding induced cardiomyopathy, which is characterized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service