Skip to Content
Merck
All Photos(2)

Documents

S9305

Sigma-Aldrich

SYBR® Green II RNA gel stain

greener alternative

10,000 × in DMSO

Synonym(s):

RNA gel dye, SYBR® RNA dye, safer gel stain

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.52

usage

1 mL sufficient for 100 mini-gels

Quality Level

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

10,000 × in DMSO

greener alternative category

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

SYBR® Green II is a highly sensitive stain for post electrophoresis staining of RNA and ssDNA in agarose or polyacrylamide gels. SYBR® Green II is not selective for RNA staining but does exhibit a higher quantum yield when bound to RNA than to double stranded DNA.
Sigma Life Science is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has inherently safer chemistry, compared to the standard use of ethidium bromide for staining. For more information see publication found in related content.

Application

SYBR® Green II RNA gel stain has been used:

  • for the quantification of ribonucleic acid (RNA) in Escherichia coli
  • for staining 5′-ETS rRNA species separated on polyacrylamide gel prior to Northern blot analysis
  • to stain formaldehyde agarose gels to visualize RNA bands

Features and Benefits

  • Ultrasensitive stain for post-electrophoresis staining of RNA and ssDNA
  • Excited maximally at 497 nm with a secondary excitation peak at 254nm
  • The fluorescence emission of SYBR®Green II stained RNA is centered at 520 nm
  • Staining agarose/formaldehyde gels with SYBR Green II does not interfere with the transfer of RNA to membranes or subsequent hybridization in Northern blot analysis as long as 0.1% -0.3% SDS is included in prehybridization and hybridization buffers to remove the dye
  • Might facilitate the detection of viroid RNAs and multicopy cellular RNA species
  • More sensitive than ethidium bromide in applications that require extremely sensitive detection techniques such as single-strand conformation polymorphism (SSCP) analysis

Legal Information

SYBR is a registered trademark of Life Technologies

replaced by

Product No.
Description
Pricing

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human ??defensin-1 and -2 expression in the gingiva of patients with specific periodontal diseases
S. Vardar-Sengul
Journal of Periodontal Research (2007)
Benjamin Lau et al.
Molecular cell, 81(2), 293-303 (2020-12-17)
Ribosome assembly is catalyzed by numerous trans-acting factors and coupled with irreversible pre-rRNA processing, driving the pathway toward mature ribosomal subunits. One decisive step early in this progression is removal of the 5' external transcribed spacer (5'-ETS), an RNA extension
Michael Richmond et al.
Biochemistry, 50(12), 2298-2312 (2011-02-09)
In this work, we describe RapA-dependent polyadenylation of model RNA substrates and endogenous, RNA polymerase-associated nucleic acid fragments. We demonstrate that the Escherichia coli RNA polymerase obtained through the classic purification procedure carries endogenous RNA oligonucleotides, which, in the presence
Dyes for in vitro detection of nucleic acids and proteins.
BioProbes, 20, 12-13 (1994)
L G Lee et al.
Cytometry, 7(6), 508-517 (1986-11-01)
The purpose of this study was to find a 488-nm excitable fluorescent dye for reticulocyte analysis by the use of fluorescence activated cell cytometry. The chemical structure of thioflavin T, a dye used for reticulocyte analysis with 457-nm excitation, was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service