Skip to Content
Merck
All Photos(2)

Key Documents

P5744

Sigma-Aldrich

Phosphatase substrate

powder

Synonym(s):

4-Nitrophenyl phosphate disodium salt hexahydrate, pNPP disodium salt hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OP(O)(ONa)2 · 6H2O
CAS Number:
Molecular Weight:
371.14
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83
Pricing and availability is not currently available.

Product Name

Phosphatase substrate, 40 mg capsules

form

powder

storage temp.

−20°C

SMILES string

O.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)c1ccc(OP([O-])([O-])=O)cc1

InChI

1S/C6H6NO6P.2Na.6H2O/c8-7(9)5-1-3-6(4-2-5)13-14(10,11)12;;;;;;;;/h1-4H,(H2,10,11,12);;;6*1H2/q;2*+1;;;;;;/p-2

InChI key

KAKKHKRHCKCAGH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Chromogenic substrate for use with alkaline and acid phosphatase ELISA procedures. Produces a soluble end product that is yellow in color and can be read spectrophotometrically at 405 nm. The pNPP reaction may be stopped with 3 N NaOH and read at 405 nm.

Other Notes

Substrate for the determination of acid and alkaline phosphatases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth A Shirtcliff et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(8), 2963-2967 (2009-02-04)
It is well known that children need solicitous parenting and a nurturing rearing environment to ensure their normal behavioral development. Early adversity often negatively impacts emotional and mental well-being, but it is less clearly established how much the maturation and
Ahmed G Abu-Shahba et al.
Tissue engineering and regenerative medicine, 17(4), 477-493 (2020-05-26)
Insufficient vascularization hampers bone tissue engineering strategies for reconstructing large bone defects. Delivery of prolyl-hydroxylase inhibitors (PHIs) is an interesting approach to upregulate vascular endothelial growth factor (VEGF) by mimicking hypoxic stabilization of hypoxia-inducible factor-1alpha (HIF-1α). This study assessed two
Christina Papista et al.
Kidney international, 88(2), 276-285 (2015-03-26)
IgA1 complexes containing deglycosylated IgA1, IgG autoantibodies, and a soluble form of the IgA receptor (sCD89), are hallmarks of IgA nephropathy (IgAN). Food antigens, notably gluten, are associated with increased mucosal response and IgAN onset, but their implication in the
Masaya Yokota et al.
Arthritis research & therapy, 16(2), R72-R72 (2014-03-19)
In addition to the pivotal roles of mast cells in allergic diseases, recent data suggest that mast cells play crucial roles in a variety of autoimmune responses. However, their roles in the pathogenesis of autoimmune skeletal muscle diseases have not
Jörn Karhausen et al.
Critical care medicine, 41(9), e200-e210 (2013-03-13)
Cardiac surgery, especially when employing cardiopulmonary bypass and deep hypothermic circulatory arrest, is associated with systemic inflammatory responses that significantly affect morbidity and mortality. Intestinal perfusion abnormalities have been implicated in such responses, but the mechanisms linking local injury and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service