Skip to Content
Merck
All Photos(2)

Documents

N2765

Sigma-Aldrich

4-Nitrophenyl phosphate disodium salt hexahydrate

chromogenic, tablet

Synonym(s):

p-Nitrophenyl phosphate disodium hexahydrate, Disodium 4-nitrophenyl phosphate hexahydrate, di-Sodium 4-nitrophenyl phosphate, pNPP disodium hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OP(O)(ONa)2 · 6H2O
CAS Number:
Molecular Weight:
371.14
Beilstein:
5324661
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

4-Nitrophenyl phosphate disodium salt hexahydrate, tablet

form

tablet

solubility

water, high purity: 0.25 tablet/mL, clear, colorless

storage temp.

−20°C

SMILES string

O.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)c1ccc(OP([O-])([O-])=O)cc1

InChI

1S/C6H6NO6P.2Na.6H2O/c8-7(9)5-1-3-6(4-2-5)13-14(10,11)12;;;;;;;;/h1-4H,(H2,10,11,12);;;6*1H2/q;2*+1;;;;;;/p-2

InChI key

KAKKHKRHCKCAGH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Chromogenic substrate for the determination of acid and alkaline phosphatases. The reaction produces a soluble end product that is yellow in color. The reaction may be stopped with 3 N NaOH and read at 405 nm.

Application

4-Nitrophenyl phosphate disodium salt hexahydrate has been used:
  • as a substrate for alkaline phosphatase coupled anti-rabbit antibody in enzyme-linked immunosorbent assay (ELISA)
  • as a substrate for the mouse secondary antibody linked to alkaline phosphatase in immunoenzymometric assay
  • in ELISA to analyze the presence of antibodies to Pseudomonas aeruginosa in animal serum sample

Quantity

Contains 20 mg substrate per tablet

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda L Miller et al.
PLoS pathogens, 16(7), e1008591-e1008591 (2020-07-10)
Reactive arthritis, an autoimmune disorder, occurs following gastrointestinal infection with invasive enteric pathogens, such as Salmonella enterica. Curli, an extracellular, bacterial amyloid with cross beta-sheet structure can trigger inflammatory responses by stimulating pattern recognition receptors. Here we show that S.
Marcus J Hines et al.
Cell reports, 33(9), 108436-108436 (2020-12-03)
The phosphatidylinositol 3-kinase (PI3K) signaling cascade downstream of the B cell receptor (BCR) signalosome is essential for B cell maturation. Proper signaling strength is maintained through the PI3K negative regulator phosphatase and tensin homolog (PTEN). Although a role for microRNA
Mihaela Gadjeva et al.
Journal of immunology (Baltimore, Md. : 1950), 184(1), 296-302 (2009-12-02)
The inflammatory response to Pseudomonas aeruginosa is not properly regulated in the lungs of patients with cystic fibrosis (CF). In the lung epithelium of individuals with wild-type CF transmembrane conductance regulator, lipid rafts containing CF transmembrane conductance regulator are rapidly
Masaya Yokota et al.
Arthritis research & therapy, 16(2), R72-R72 (2014-03-19)
In addition to the pivotal roles of mast cells in allergic diseases, recent data suggest that mast cells play crucial roles in a variety of autoimmune responses. However, their roles in the pathogenesis of autoimmune skeletal muscle diseases have not
Christina Papista et al.
Kidney international, 88(2), 276-285 (2015-03-26)
IgA1 complexes containing deglycosylated IgA1, IgG autoantibodies, and a soluble form of the IgA receptor (sCD89), are hallmarks of IgA nephropathy (IgAN). Food antigens, notably gluten, are associated with increased mucosal response and IgAN onset, but their implication in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service