Skip to Content
Merck
All Photos(6)

Key Documents

TRANSRTRO

Roche

Transcriptor Reverse Transcriptase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106313
NACRES:
NA.21

recombinant

expressed in E. coli

Quality Level

Assay

≥90% (SDS-PAGE)

form

liquid

usage

sufficient for 25 reactions
sufficient for 50 reactions
sufficient for 500 reactions

specific activity

0.05 U/mg

feature

dNTPs included: no
hotstart: no

manufacturer/tradename

Roche

packaging

pkg of 200 reactions (03531287001)
pkg of 25 reactions (03531317001)
pkg of 50 reactions (03531295001)

technique(s)

RT-PCR: suitable
RT-qPCR: suitable

color

colorless

pH

7.2

solubility

water: miscible

suitability

suitable for molecular biology

UniProt accession no.

application(s)

genomic analysis
life science and biopharma

detection method

probe-based

foreign activity

DNase activity, none detected
Nicking activity, none detected
RNase activity, none detected

storage temp.

−20°C (−15°C to −25°C)

General description

A recombinant reverse transcriptase for the robust transcription of RNA fragments up to 14 kb used in two-step RT-PCR on thermal cyclers and real-time PCR instruments.
In retroviruses like the human immunodeficiency virus type 1 (HIV-1), reverse transcriptase (RT) is the core enzyme. HIV-1 RT is made of two subunits of 66 kDa and 51 kDa (p66 and p5l). A human endogenous retrovirus of the HERV-K family codes for reverse transcriptase (RT) enzyme.

Application

Transcriptor Reverse Transcriptase is designed to transcribe RNA (mRNA, total RNA, viral RNA, and in vitro-transcribed RNA) from a variety of sources, using conventional thermal cyclers and real-time PCR instruments (e.g., the LightCycler® Instruments) for the following applications:

  • Synthesis of first-strand cDNA for use in subsequent amplification reactions
  • RT-PCR of GC-rich RNA templates
  • Cy3, Cy5, DIG, biotin, and aminoallyl labeling during cDNA synthesis
  • Retrieving and cloning the 5′ and 3′ termini of mRNA by RACE
  • Generation of cDNA libraries with large inserts
  • Dideoxy DNA sequencing
  • RNA sequencing
  • 3′-end labeling of DNA fragments
  • Generation of single-stranded probes for genomic footprints
  • In the reverse transcription of RNA from human Papillomavirus E6, cortical and striatal tissues, muscle biopsies of Becker muscular dystrophy samples and miRNA stem-loop from oocytes and human cerebral microvascular endothelial cells (hCMVECs)

Biochem/physiol Actions

Human immunodeficiency virus type 1 reverse transcriptase (HIV-1 RT) is essential for the catalytic conversion of single-stranded viral RNA into the double-stranded linear DNA that is integrated into host cell chromosomes.

Features and Benefits

  • Achieve high sensitivity in two-step RT-PCR.
Transcriptor Reverse Transcriptase is used in conventional thermal cyclers and real-time PCR instruments (e.g., the LightCycler®Instruments).
  • Obtain more full-length transcripts - up to 14 kb.
cDNA libraries with large inserts can be generated.
  • Reverse transcribe difficult templates.
The enzyme works well at elevated temperatures, thereby overcoming RNA secondary structure (e.g., GC-rich RNA templates) and facilitating optimal reaction conditions.
  • Efficiently label cDNA.
Cy3-, Cy5-, DIG-, biotin-, or aminoallyl-labeled nucleotides are incorporated during cDNA synthesis.

Components

  • Transcriptor Reverse Transcriptase, in storage buffer
  • Transcriptor RT buffer, 5x concentrated

Quality

Each lot of Transcriptor Reverse Transcriptase is routinely function tested in RT-PCR:

  • with a conventional thermal cycler to detect a 10-kb fragment from the human dystrophin gene, starting from human skeletal muscle total RNA
  • with the LightCycler® Instrument to detect 5 x 102 to 5 x 106 copies of in vitro-transcribed human PBGD RNA. The results are defined in fixed crossing points and fixed fluorescence intensity.

Preparation Note

Volume activity: 20 U/μl.
Transcription of long, rare, or difficult RNA targets
Transcriptor Reverse Transcriptase is recommended for RT-PCR of:

  • long targets, because it can transcribe up to 14 kb RNA templates
  • rare targets, because it has high sensitivity
  • GC-rich targets, because it can operate at high temperatures (up to +65oC) to eliminate problems associated with extensive secondary structure

Labeling for many applications
Transcriptor Reverse Transcriptase is also recommended for preparing labeled cDNA, since it accepts a wide variety of modified nucleotides (including Cy3-, Cy5-, DIG-, Biotin-, or aminoallyl-labeled dNTPs).
Reaction requirements
Transcriptor accepts ssRNA and ssDNA templates and requires a primer for transcription.

Storage and Stability

Avoid repeated freezing and thawing.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

LightCycler is a registered trademark of Roche

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pietro Spitali et al.
Journal of cachexia, sarcopenia and muscle, 9(4), 715-726 (2018-04-24)
Analysis of muscle biopsies allowed to characterize the pathophysiological changes of Duchenne and Becker muscular dystrophies (D/BMD) leading to the clinical phenotype. Muscle tissue is often investigated during interventional dose finding studies to show in situ proof of concept and
Expression profiles and function analysis of microRNAs in postovulatory aging mouse oocytes
Wang TY, et al.
Aging, 9, 1186-1186 (2017)
Crystal structure of human immunodeficiency virus type 1 reverse transcriptase complexed with double-stranded DNA at 3.0 A resolution shows bent DNA.
Jacobo-Molina A, et al.
Proceedings of the National Academy of Sciences of the USA, 90(13), 6320-6324 (1993)
Age-and sex-related changes in cortical and striatal nitric oxide synthase in the Q175 mouse model of Huntington's disease
Padovan-Neto FE, et al.
Nitric Oxide, 83, 40-50 (2019)
Identification of an active reverse transcriptase enzyme encoded by a human endogenous HERV-K retrovirus.
Berkhout B, et al.
Journal of Virology, 73(3), 2365-2375 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service