N9389
4-Nitrophenyl phosphate disodium salt hexahydrate
chromogenic, tablet
Synonym(s):
p-Nitrophenyl phosphate disodium hexahydrate, Disodium 4-nitrophenyl phosphate hexahydrate, di-Sodium 4-nitrophenyl phosphate, pNPP disodium hexahydrate
About This Item
Recommended Products
product name
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
description
5 mg substrate per tablet
form
tablet
fluorescence
λex 310 nm (pH 7.1)
λex 399 nm (Reaction Product)
storage temp.
−20°C
SMILES string
O.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)c1ccc(OP([O-])([O-])=O)cc1
InChI
1S/C6H6NO6P.2Na.6H2O/c8-7(9)5-1-3-6(4-2-5)13-14(10,11)12;;;;;;;;/h1-4H,(H2,10,11,12);;;6*1H2/q;2*+1;;;;;;/p-2
InChI key
KAKKHKRHCKCAGH-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
Application
- enzyme-linked immunosorbent assay (ELISA) to visualize glycoprotein VI (GPVI) binding to collagen type I (Col I)
- enzymatic color reaction for semi-quantitative measurement of allergen-specific IgE by ELISA
- alkaline phosphatase assay
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service