Skip to Content
Merck
All Photos(1)

Documents

P0114

Sigma-Aldrich

Phosphatase, Alkaline from bovine intestinal mucosa

BioUltra, ≥5,700 DEA units/mg protein

Synonym(s):

Alkaline phosphatase, Orthophosphoric-monoester phosphohydrolase (alkaline optimum)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
42010105
NACRES:
NA.54

product line

BioUltra

form

(40% glycerol containing 6 mM Tris, 6 mM MgCl2 and 0.12 mM ZnCl2, pH approximately 7.6)

specific activity

≥5,700 DEA units/mg protein

mol wt

dimer ~160 kDa

concentration

≥10 mg/mL

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Alkaline phosphatase from bovine intestinal mucosa is both a phosphomonoesterase and a pyrophosphatase.

Application

Alkaline phosphatase can be used to dephosphorylate casein and other proteins. Alkaline phosphatase may be also be used to dephosphorylate the 5′-termini of DNA or RNA to prevent self-ligation. DNA or RNA can also be tagged with radiolabeled phosphate (via T4 polynucleotide kinase) after dephosphorylation with alkaline phosphatase..
High specific activity grade recommended for antibody and protein conjugation.
Phosphatase alkaline from bovine intestinal mucosa has been used in a study to measure the hydrolysis kinetics of three polyfluorinated alkyl phosphate monoesters, differing in fluorinated chain length. It has also been used in a study to show that the developmentally controlled expression of the isosomes C-1 and C-2 indicate functional changes in intestinal alkaline phosphatase with tissue maturation.

Biochem/physiol Actions

Alkaline phosphatase was shown to have anti-inflammatory effects and can reduce the severity of septic shock by detoxifying lipid A.
The enzyme has a broad specificity for phosphate esters of alcohols, amines, pyrophosphate, and phenols. It is routinely used to dephosphorylate proteins and nucleic acids.

Physical properties

Bovine intestinal alkaline phosphatase is a dimeric, membrane-derived glycoprotein. At least three isoforms exist, which typically possess two N-linked and one or more O-linked glycans per monomer.
Bovine intestinal alkaline phosphatase is a dimeric, membrane-derived glycoprotein. At least three isoforms exist, which typically possess two N-linked and one or more O-linked glycans per monomer.2 The enzyme requires zinc, and magnesium or calcium divalent ions for activity.

Unit Definition

One DEA unit will hydrolyze 1 μmole of 4-nitrophenyl phosphate per minute at pH 9.8 at 37 °C. (One glycine unit is equivalent to ~3 DEA units)

Physical form

Solution in 40% glycerol containing 6 mM Tris, 6 mM MgCl2 and 0.12 mM ZnCl2, pH approximately 7.6

Analysis Note

Package sizes are based on DEA units

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dagmara Wojcik-Grzybek et al.
International journal of molecular sciences, 23(6) (2022-03-26)
Inflammatory bowel diseases (IBD) are commonly considered as Crohn's disease and ulcerative colitis, but the possibility that the alterations in gut microbiota and oxidative stress may affect the course of experimental colitis in obese physically exercising mice treated with the
M Besman et al.
The Journal of biological chemistry, 260(20), 11190-11193 (1985-09-15)
Alkaline phosphatases from calf and bovine small intestines have been isolated in homogeneous form from both mucosa and luminal contents. The detergent-solubilized calf enzyme resolves into two peaks of activity, C-1 and C-2, on chromatofocusing. Only one of these activity
Fuhong Su et al.
Critical care medicine, 34(8), 2182-2187 (2006-06-16)
Alkaline phosphatase may decrease the harmful effects of lipopolysaccharide by detoxifying lipid A. The aim of this study was to investigate whether administration of alkaline phosphatase is beneficial in a clinically relevant septic shock model. Interventional laboratory study. University hospital
Suzanne Kats et al.
Recent patents on inflammation & allergy drug discovery, 3(3), 214-220 (2009-06-19)
Laboratory and clinical data have implicated endotoxin as an important factor in the inflammatory response to cardiopulmonary bypass. Alkaline phosphatase prevents endotoxin-induced systemic inflammation in animals and humans. We assessed the effects of the administration of bovine intestinal alkaline phosphatase
Derek A Jackson et al.
Environmental toxicology and chemistry, 31(9), 1966-1971 (2012-06-21)
The hydrolysis kinetics of three polyfluorinated alkyl phosphate monoesters (monoPAPs), differing in fluorinated chain length, were measured using bovine intestinal alkaline phosphatase to catalyze the reaction. Kinetic values were also measured for analogous hydrogenated phosphate monoesters to elucidate the effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service