Skip to Content
Merck
All Photos(1)

Key Documents

P4557

Sigma-Aldrich

Phenol solution

Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology

Synonym(s):

Tris-equilabrated Phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O
CAS Number:
Molecular Weight:
94.11
Beilstein:
6428463
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.52
grade:
Molecular Biology
for molecular biology

grade

Molecular Biology
for molecular biology

product line

BioReagent

Assay

≥99.0% (GC)

form

liquid

dilution

(for analytical testing)

pH

7.7-8.1 (25 °C)

storage temp.

2-8°C

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

General description

A tris-equilibrated phenol solution is a clear liquid capped with a layer of Tris buffer. Phenol solutions can be used for a wide variety of molecular biology applications. Organic compounds like phenols have an aromatic carbocyclic ring with a hydroxyl group attached directly to it. The common name for monohydroxybenzene is phenol.

Application

Phenol solution has been used:

  • as a component of the digestion reaction mix to prepare RNase-free plasmids for in vitro translation in a rabbit reticulocyte lysate
  • for extraction of DNA from Arabidopsis thaliana Rosette leaves
  • in the extraction of DNA from blood on preservation paper by standard phenol-chloroform protocol
  • as a component of extraction buffer to extract RNA from fruit pulp
  • for the extraction of protein from roots
  • for the isolation of fecal DNA
  • for the extraction of caged RNA

Biochem/physiol Actions

The phenol-chloroform extraction method is widely employed to extract nucleic acids. Even though phenol does not completely inhibit RNase activity, its unique characteristic makes it beneficial for the isolation of nucleic acids when used in combination with chloroform and alcohol.

Components

  • 1 bottle Tris-equilabrated Phenol (product number P4557)
  • Equilibration Buffer (product B5658)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nucleic Acid Extraction Techniques
Yi-Wei Tang and Charles W. Stratton
Journal of Separation Science (2012)
Fecal Microbiome and Food Allergy in Pediatric Atopic Dermatitis: A Cross-Sectional Pilot Study
Fieten KB, et al.
International Archives of Allergy and Immunology, 175(1-2), 77-84 (2018)
PHENOLS
Smith PWG and Tatchell AR
Aromatic Chemistry, 79-104 (1969)
Protocol Optimization of Proteomic Analysis of Korean Ginseng (Panax ginseng Meyer)
Braccia C, et al.
Journal of Separation Science, 8(4), 53-53 (2021)
Tim J Bull et al.
Journal of clinical microbiology, 41(7), 2915-2923 (2003-07-05)
Mycobacterium avium subsp. paratuberculosis is a robust and phenotypically versatile pathogen which causes chronic inflammation of the intestine in many species, including primates. M. avium subsp. paratuberculosis infection is widespread in domestic livestock and is present in retail pasteurized cows'

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service