C0549
Chloroform:Isoamyl alcohol 24:1
suitable for nucleic acid purification
Synonym(s):
chloroform-isoamyl alcohol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12190000
NACRES:
NA.52
application(s):
agriculture
Recommended Products
form
liquid
Quality Level
contains
0.6-1.0% ethanol as stabilizer
suitability
suitable for nucleic acid purification
application(s)
agriculture
General description
Chloroform:Isoamyl Alcohol 24:1 is an organic reagent used for DNA extraction and precipitation.
Application
Chloroform:Isoamyl alcohol 24:1 has been used:
- to extract bacterial genomic DNA, DNA from Daphnia spp, and salmon sperms.
- for a single DNA precipitation step for high-quality DNA extraction.
- for protein extraction and RNAse treatment.
- in phenolic oxidation to separate phenolics from the aqueous portion.
- for RNA isolation from brown adipose tissue (BAT), inguinal white adipose tissue (iWAT), and epididymal adipose tissue (eWAT).
- in modified cetyltrimethyl ammonium bromide (CTAB) method to extract total RNA from Impatiens balsamina.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3
Target Organs
Central nervous system, Liver,Kidney
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
185.0 °F
Flash Point(C)
85 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocol: a simple method for extracting next-generation sequencing quality genomic DNA from recalcitrant plant species
Healey A, et al.
Plant methods, 10(1), 1-8 (2014)
A modified SDS-based DNA extraction method from raw soybean
Xia YC, et al.
Bioscience Reports, 39(2), BSR20182271-BSR20182271 (2019)
Defective insulin-stimulated GLUT4 translocation in brown adipocytes induces systemic glucose homeostasis dysregulation independent of thermogenesis in female mice
Picatoste B, et al.
Molecular Metabolism, 53 (2021)
A modified CTAB-based protocol for total RNA extraction from the medicinal plant Impatiens balsamina (Balsaminaceae) for next-generation sequencing studies.
FOONG LC, et al.
Journal of Separation Science, 46(2) (2017)
Harnessing microbially generated power on the seafloor.
Tender LM
Nature Biotechnology, 20(8), 821-825 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service