Skip to Content
Merck
All Photos(1)

Key Documents

P9346

Sigma-Aldrich

Phenol solution

≥89.0% (in water)

Synonym(s):

Phenol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O
CAS Number:
Molecular Weight:
94.11
Beilstein:
6428463
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21

Product Name

Liquified Phenol, ≥89.0%

Agency

USP/NF
USP
suitable for SM 4500 - NH3

Quality Level

Assay

≥89.0%

impurities

~10% (water)

density

1.07 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

General description

Liquified Phenol is a solution of phenol containing ~10% water. It is potential as a chemical peeling agent for treating stable vitiligo and alopecia areata.

Application

Liquified Phenol has been used in a study to record the one-photon (linear) absorption (1PA) and two-photon absorption 2PA spectra of Disperse Red One (DR1). It may be used for deproteinization of cell lysate during the preparation of total nucleic acids.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S S Savant et al.
Indian journal of dermatology, venereology and leprology, 65(2), 93-98 (1999-03-01)
Chemical peeling with 88% phenol was carried out on 142 sites of stable vitiligo (hairy-126, non hairy-16) and on 69 sites of alopecia areata (AA). After cleansing and defatting, phenol was applied on affected areas till a uniform frost appeared.
J T Wachsman et al.
Journal of bacteriology, 104(2), 814-818 (1970-11-01)
Parallel studies were performed with methionineless derivatives of Escherichia coli 15 T(-) and Bacillus megaterium KM: T(-). Methylated bases are present in the total cell ribonucleic acid (RNA) of B. megaterium. The level of RNA methylation in E. coli is
Untangling the excited states of DR1 in solution: An experimental and theoretical study.
De Boni L, et al.
The Journal of Physical Chemistry A, 112(17), 3886-3890 (2008)
Thitiporn Anunthawan et al.
Biochimica et biophysica acta, 1848(6), 1352-1358 (2015-03-15)
We investigated the mechanisms of two tryptophan-rich antibacterial peptides (KT2 and RT2) obtained in a previous optimization screen for increased killing of both Gram-negative and Gram-positive bacteria pathogens. At their minimal inhibitory concentrations (MICs), these peptides completely killed cells of
Rita A Busuttil et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(10), 2761-2772 (2014-03-25)
Gene-expression profiling has revolutionized the way we think about cancer and confers the ability to observe the synchronous expression of thousands of genes. The use of putative genome-level expression profiles has allowed biologists to observe the complex interactions of genes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service