Skip to Content
Merck
All Photos(1)

Key Documents

A2942

Sigma-Aldrich

Amphotericin B

solution, suitable for cell culture, BioReagent

Synonym(s):

Amphotericin, Fungizone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H73NO17
CAS Number:
Molecular Weight:
924.08
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.76
Pricing and availability is not currently available.

Product Name

Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture

Quality Level

sterility

0.1 μm filtered

product line

BioReagent

form

liquid

concentration

250 μg/mL in deionized water

technique(s)

cell culture | mammalian: suitable

color

yellow

antibiotic activity spectrum

fungi
yeast

Mode of action

cell membrane | interferes

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@]12C[C@@H](O[C@@H]3O[C@H](C)[C@@H](O)[C@H](N)[C@@H]3O)\C=C\C=C\C=C\C=C\C=C\C=C\C=C\[C@H](C)[C@@H](O)[C@@H](C)[C@H](C)OC(=O)C[C@H](O)C[C@H](O)CC[C@@H](O)[C@H](O)C[C@H](O)C[C@](O)(C[C@H](O)[C@H]1C(=O)O[Na])O2

InChI

1S/C47H73NO17/c1-27-17-15-13-11-9-7-5-6-8-10-12-14-16-18-34(64-46-44(58)41(48)43(57)30(4)63-46)24-38-40(45(59)60)37(54)26-47(61,65-38)25-33(51)22-36(53)35(52)20-19-31(49)21-32(50)23-39(55)62-29(3)28(2)42(27)56/h5-18,27-38,40-44,46,49-54,56-58,61H,19-26,48H2,1-4H3,(H,59,60)/b6-5+,9-7+,10-8+,13-11+,14-12+,17-15+,18-16+/t27-,28-,29-,30+,31+,32+,33-,34-,35+,36+,37-,38-,40+,41-,42+,43+,44-,46-,47+/m0/s1

InChI key

APKFDSVGJQXUKY-INPOYWNPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amphotericin B is a macrocyclic lactone with a hydrophilic polyhydroxyl chain and a lipophilic polyene based hydrocarbon chain.[1]

Application

Amphotericin B solution has been used:
  • as an intraperitoneal injection for the treatment of Aspergillus fumigatus infection in mice[2]
  • as a constituent of aspiration medium for the culture of Chinese Merino ewe ovaries
  • as a constituent of transport medium during the collection of equine corpora lutea[3]
  • as a supplement in RPMI medium for the culture of the mouse-mouse B cell hybridoma 55-6 cell line[4]

Biochem/physiol Actions

Amphotericin B is a polyene antifungal antibiotic from Streptomyces sp. that is clinically used to treat systemic fungal infections.[5] It has a high affinity for sterols, especially ergosterols, of both fungal and bacterial cell membranes. Normal usage in maintenance of antifungal cell cultures is 2.5 mg/L with penicillin and streptomycin used in the medium.
Amphotericin B is most effective against fungi and yeast.[6] It forms transmembrane channels by binding to sterols associated with membranes, thereby leading to the leakage of small molecules.[6] Amphotericin B elicits toxicity despite being an effective antibiotic for treating mycoses.[7]

Packaging

20,50,100ml

Components

This product is supplied as 250 μg/mL Amphotericin B in deionized water. It is a solution containing Amphotericin B, and the following components aimed to improve its solubility: sodium deoxycholate, sodium chloride and sodium phosphate.

Caution

Amphotericin B remains active for 3 days in culture at 37°C.

Preparation Note

It is not unusual for this material to form a precipitate in aqueous solutions. Mixing the solution will redissolve most of the precipitate but, if it does not fully resolubilize, the Amphotericin B will still be suitable for use.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Light sensitive.

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Lourdes Acosta et al.
Cytotechnology, 54(3), 189-200 (2008-11-13)
Batch cultures were carried out to study the kinetic, stoichiometry, and regulation of glucose and glutamine metabolism of a murine hybridoma line. Asymmetric logistic equations (ALEs) were used to fit total and viable cell density, and nutrient and metabolite/product concentrations.
Yong-Hong Wang et al.
The Journal of antibiotics, 63(9), 553-557 (2010-07-08)
A new derivative of amphotericin B named amphotericin B(5) has been obtained under the following condition: amphotericin B powder was first dissolved in N,N-dimethylformamide at room temperature to obtain a solution of concentration of 2.0 mg ml(-1). Then, the solution
J Bolard et al.
Biochemistry, 30(23), 5707-5715 (1991-06-11)
The inducement of K+ permeability through membranes by the polyene antibiotic amphotericin B (AmB) has been analyzed as a measure of the antibiotic activity. Dose-response curves have been obtained with cholesterol- and ergosterol-containing egg yolk phosphatidylcholine large unilamellar vesicles (LUVs)
Sarah C Brüningk et al.
Scientific reports, 10(1), 1653-1653 (2020-02-06)
For multimodality therapies such as the combination of hyperthermia and radiation, quantification of biological effects is key for dose prescription and response prediction. Tumour spheroids have a microenvironment that more closely resembles that of tumours in vivo and may thus
Ewa Grela et al.
Scientific reports, 9(1), 17029-17029 (2019-11-21)
Amphotericin B is an antibiotic used as the "gold standard" in the treatment of life-threatening fungal infections. Several molecular mechanisms have been proposed to explain exceptionally high effectiveness of amphotericin B in combating fungi. In the present work, we apply

Questions

1–4 of 4 Questions  
  1. At what concentration should Product A2942, Amphotericin B solution, be used?

    1 answer
    1. We suggest a working concentration of 10 ml/L (2.5 mg/L).

      Helpful?

  2. What is the shelf life of Product A2942, Amphotericin B solution?

    1 answer
    1. It has an expiration date of 18 months. This can be found on the lot-specific certificate of analysis.

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. How should Product A2942, Amphotericin B solution, be stored?

    1 answer
    1. This product should be stored at -20 °C.  It is stable in culture at 37 °C up to 3 days.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service