Skip to Content
Products
Cart
0
AD
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Magnesium chloride solution
Linear Formula:
MgCl
2
CAS Number:
7786-30-3
Molecular Weight:
95.21
Products
Product Category
salts (4)
Brand
Sigma-Aldrich (2)
Color
Formula Weight
pH Value
Physical Form
Product Line
Research Area
Quality Segment
Application
Package Size
Sterility
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
M1028
Magnesium chloride solution,
Molecular Biology, 1.00 M±0.01 M
Expand
View Pricing
68475
Magnesium chloride solution,
BioUltra, Molecular Biology, 2 M in H
2
O
Expand
View Pricing
M8787
Magnesium chloride solution,
PCR Reagent, 25 mM MgCI
2
solution for PCR
Expand
View Pricing
00457
Magnesium chloride solution,
0.1 M
Expand
View Pricing
63069
Magnesium chloride solution,
BioUltra, Molecular Biology, ~1 M in H
2
O
Expand
View Pricing
63036
Magnesium chloride solution,
BioUltra, Molecular Biology, ~0.025 M in H
2
O
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Español - ES
Français - FR
Learn More