Skip to Content
Merck
All Photos(1)

Key Documents

M7403

Sigma-Aldrich

MCDB 153 Medium

With trace elements, L-glutamine and 28mM HEPES, without sodium bicarbonate, powder, suitable for cell culture

Synonym(s):

Molecular, Cellular, and Development Biology 153 medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

components

NaHCO3: no
L-glutamine: 0.8772 g/L
glucose: 1.081 g/L (Dextro)
sodium pyruvate: 0.055 g/L
phenol red: 0.001242 g/L
HEPES: 6.6 g/L (25mM)

shipped in

ambient

storage temp.

2-8°C

General description

MCDB 153 media are modifications of Ham′s nutrient mixture F-12, designed for the growth of human keratinocytes, clonal growth of chicken embryo fibroblasts, and Chinese hamster ovary (CHO) cells using low levels of fetal bovine serum protein (FBSP), extensive trace elements or no serum protein.

Application

MCDB 153 Medium has been used as a component of melanoma isolation media (MIM) to culture human non-metastatic and metastatic melanoma cells.

Quantity

Formulated to contain 17.6 grams of powder per liter of medium.

Reconstitution

Supplement with 1.18 g/L sodium bicarbonate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hans Peter Weitzenböck et al.
Cancer medicine, 11(4), 956-967 (2021-12-25)
Malignant melanoma is the deadliest form of skin cancer and NRF2 has been proposed as a main regulator of tumor cell malignancy. Still the mechanisms how NRF2 is contributing to melanoma progression are incompletely understood. Here we analyzed the effects
Andrew S McNeal et al.
eLife, 10 (2021-11-24)
Benign melanocytic nevi frequently emerge when an acquired BRAFV600E mutation triggers unchecked proliferation and subsequent arrest in melanocytes. Recent observations have challenged the role of oncogene-induced senescence in melanocytic nevus formation, necessitating investigations into alternative mechanisms for the establishment and
Matthew L Fisher et al.
Molecular carcinogenesis, 55(12), 2024-2036 (2015-12-24)
Melanoma is a metastatic cancer associated with poor survival. Here, we study a subpopulation of melanoma cancer cells displaying melanoma cancer stem cell (MCS cells) properties including elevated expression of stem cell markers, increased ability to survive as spheroids, and
Dominik Brokatzky et al.
The EMBO journal, 38(11) (2019-04-14)
Apoptosis is a frequent form of programmed cell death, but the apoptotic signaling pathway can also be engaged at a low level, in the absence of cell death. We here report that such sub-lethal engagement of mitochondrial apoptosis signaling causes
Yochai Wolf et al.
Cell, 179(1), 219-235 (2019-09-17)
Although clonal neo-antigen burden is associated with improved response to immune therapy, the functional basis for this remains unclear. Here we study this question in a novel controlled mouse melanoma model that enables us to explore the effects of intra-tumor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service