Skip to Content
Merck
All Photos(1)

Key Documents

C8052

Sigma-Aldrich

Cholera Toxin

from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder, A-B type toxin

Synonym(s):

CTX, Cholera enterotoxin, Choleragen

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Cholera Toxin from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder

Quality Level

Assay

≥90% (SDS-PAGE)

form

lyophilized powder

composition

Protein, ~5% Lowry-TCA

solubility

H2O: 10 mg/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Cholera toxin is a member of the AB5-subunit family of toxins. It consists of a single A subunit responsible for ADP-ribosylation and five B subunits arranged as a pentameric ring with cell surface receptor binding and internalization functions. The A subunit catalyzes the ADP-ribosylation of the α-subunit of the stimulatory G protein (Gαs), leading to increased adenylyl cyclase activity and cAMP levels. It also ADP-ribosylates transducing and tubulin.

Cholera toxin and its A subunit are commonly used to study signal transduction mechanisms due to their impact on adenylate cyclase. Additionally, cholera toxin acts as an adjuvant by stimulating B lymphocytes and T helper cell type 2 responses by inhibiting interleukin-12 production. The non-toxic B subunit (CTB) attaches to cells by binding to ganglioside GM1, making it a useful label for microglial cells in neurological research. It has also proved to be an excellent tracer for axonal transport studies using immunohistochemical methods.

Application

Cholera Toxin from Vibrio cholerae has been used as a positive control in cAMP (cyclic AMP) assay for enterotoxins. It has been added as a supplement in cell culture media of primary tumors and epithelial cells.
Cholera Toxin from Vibrio cholerae has been used in toxin neutralization assays to assess the cytotoxicity of cells.

Biochem/physiol Actions

Toxin consisting of an A subunit (27 kDa) surrounded by five B subunits (approximately 12 kDa each), which attach the toxin to ganglioside GM1 on the cell surface.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Packaging

Package size based on protein content.

Physical form

Lyophilized powder containing Tris buffer salts, sodium chloride, sodium azide and sodium EDTA

Reconstitution

When reconstituted at 1 mg/mL in water, solution will contain 0.05 M Tris buffer salts, pH 7.5, 0.2 M NaCl, 0.003 M NaN3, and 0.001 M sodium EDTA. Store reconstituted solutions in the refrigerator.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 3

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A recombinant probiotic for treatment and prevention of cholera
Focareta A, et al.
Gastroenterology, 130, 1688-1695 (2006)
Rapid Expansion of Human Epithelial Stem Cells Suitable for Airway Tissue Engineering.
Butler CR et al.
American Journal of Respiratory and Critical Care Medicine, 194, 156-156 (2016)
Cholera toxin subunit B as adjuvant--an accelerator in protective immunity and a break in autoimmunity
Stratmann T
Vaccines, 3, 579-596 (2015)
Structure, biological functions and applications of the AB5 toxins
Beddoe T, et al.
Trends in Biochemical Sciences, 35, 411-418 (2010)
Enhanced degradation of stimulatory G-protein (Gsalpha) by cholera toxin is mediated by ADP-ribosylation of Gsalpha protein but not by increased cyclic AMP levels
Shah BH
Advances in Experimental Medicine and Biology, 93-97 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service