Skip to Content
Merck
All Photos(1)

Key Documents

L4386

Sigma-Aldrich

L-15 Medium (Leibovitz)

With ʟ-glutamine and sodium pyruvate, powder, suitable for cell culture

Synonym(s):

Leibovitz L15

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Product Name

L-15 Medium (Leibovitz), With L-glutamine, powder, suitable for cell culture

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

sodium pyruvate: 0.55 g/L
phenol red: 0.011 g/L
L-glutamine: 0.3 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

L-15 Medium (Leibovitz) is formulated for use in carbon dioxide (CO2) free systems requiring sodium bicarbonate supplements. It supports established cell lines, such as human larynx epidermoid cells (HEp-2) and Rhesus monkey kidney epithelial cells (LLC-MK2), as well as primary explants of embryonic and adult human tissue. L-15 medium can also be used for culturing viruses.

Application

L-15 Medium (Leibovitz) has been used:
  • to study the toxicity of silver nanoparticles (AgNPs) with primary cultures of Eisenia fetida coelomocytes
  • to culture crab hepatopancreas and ovary tissues
  • as a component of the dissociation medium for infusion of the pancreas of mice

Quantity

Formulated to contain 13.8 grams of powder per liter of medium.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Qian et al.
Insect biochemistry and molecular biology, 27(5), 387-395 (1997-05-01)
Prostaglandins of the 2-series (e.g. PGE2) are typically synthesized from arachidonic acid (AA) after AA is released from cellular phospholipids after activation of an intracellular phospholipase A2 (PLA2). Treatment of isolated salivary glands with PLA2 inhibitor oleyloxyethyl phosphorylcholine (OPC) or
Effect of estradiol on vitellogenesis and oocyte development of female swimming crab, Portunus trituberculatus
Liu M, et al.
Aquaculture (Amsterdam, Netherlands), 486, 240-245 (2018)
Sarah Johann et al.
Toxics, 8(2) (2020-07-02)
Genotoxicity assessment is of high relevance for crude and refined petroleum products, since oil compounds are known to cause DNA damage with severe consequences for aquatic biota as demonstrated in long-term monitoring studies. This study aimed at the optimization and
Steven J Conrad et al.
Avian diseases, 64(3), 243-246 (2020-11-19)
Marek's disease (MD) is an oncogenic, lymphoproliferative, and highly contagious disease of chickens. Its etiologic agent is the alphaherpesvirus Marek's disease virus (MDV, Gallid alphaherpesvirus 2), and it is a chronic and ubiquitous problem for the poultry industry with significant
Joshua Phelan et al.
Talanta, 211, 120737-120737 (2020-02-20)
This is the first demonstration of the study of glycan protein turnover in living cells by FTIR with commercially available tetraacetylated N-Azidoacetyl-D-Mannosamine (Ac4ManNAz) label. The FTIR analysis has shown to be able to monitor the metabolism of glycans in living

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service