Skip to Content
Merck
All Photos(1)

Key Documents

I0516

Sigma-Aldrich

Insulin solution from bovine pancreas

10 mg/mL insulin in 25  mM HEPES, pH 8.2, BioReagent, sterile-filtered, suitable for cell culture

Synonym(s):

Bovine insulin, Cell culture grade insulin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.77
biological source:
bovine
form:
solution

biological source

bovine

sterility

sterile-filtered

product line

BioReagent

form

solution

concentration

10 mg/mL insulin in 25  mM HEPES, pH 8.2

technique(s)

cell culture | mammalian: suitable
single cell analysis: suitable

impurities

endotoxin, tested

pH

7.9-8.5

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

SMILES string

SC[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CN)[C@H](CC)C)C(C)C)CCC(=O)O)CCC(=O)N)CS)C(=O)N[C@@H](C)C(=O)N[C@@H](CO)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CS)C(=O)N[C@@H](CO)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](Cc4ccc(cc4)O)C(=O)N[

InChI

1S/2C97H151N25O34S4/c2*1-12-46(10)77(120-72(131)35-98)96(154)122-76(45(8)9)94(152)107-55(24-28-74(134)135)79(137)104-53(22-26-69(100)128)82(140)116-66(40-159)93(151)118-64(38-157)90(148)103-47(11)78(136)114-63(37-124)89(147)121-75(44(6)7)95(153)119-67(41-160)92(150)115-62(36-123)88(146)109-57(30-43(4)5)84(142)110-58(31-48-13-17-50(125)18-14-48)85(143)105-52(21-25-68(99)127)80(138)108-56(29-42(2)3)83(141)106-54(23-27-73(132)133)81(139)112-60(33-70(101)129)87(145)111-59(32-49-15-19-51(126)20-16-49)86(144)117-65(39-158)91(149)113-61(97(155)156)34-71(102)130/h2*13-20,42-47,52-67,75-77,123-126,157-160H,12,21-41,98H2,1-11H3,(H2,99,127)(H2,100,128)(H2,101,129)(H2,102,130)(H,103,148)(H,104,137)(H,105,143)(H,106,141)(H,107,152)(H,108,138)(H,109,146)(H,110,142)(H,111,145)(H,112,139)(H,113,149)(H,114,136)(H,115,150)(H,116,140)(H,117,144)(H,118,151)(H,119,153)(H,120,131)(H,121,147)(H,122,154)(H,132,133)(H,134,135)(H,155,156)/t2*46-,47-,52-,53-,54-,55-,56-,57-,58-,59-,60-,61-,62-,63-,64-,65-,66-,67-,75-,76-,77-/m00/s1

InChI key

XYPANFGXFLOTDK-CJLNJPJHSA-N

Gene Information

bovine ... INS(280829)
cow ... INS(280829)

Application

Insulin solution from bovine pancreas has been used:
  • to study the effect of exogenous insulin on plasma concentrations of glucose, insulin, insulin-like growth factor I (IGF-I), and progesterone (P4) in pubertal beef heifers
  • to study the insulin responsiveness of the placenta during pregnancy complications using primary human trophoblast cells and placental villous explants
  • to study the effect of insulin receptor signaling dendritic spine density in neonatal mouse hippocampal neurons in vitro

Biochem/physiol Actions

Insulin is a major glucose-lowering hormone, insufficiency of which causes hyperglycemia observed in type 2 diabetes.
Two-chain polypeptide hormone produced by the β-cells of pancreatic islets. Its molecular weight is ~5800 Da. The α and β chains are joined by two interchain disulfide bonds. The α chain contains an intrachain disulfide bond. Insulin regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids and inhibits the breakdown of glycogen, protein, and fat.

Physical form

stable 2-8 °C

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wafa B'chir et al.
Endocrinology, 159(5), 2153-2164 (2018-04-11)
Given the increasing prevalence of obesity and the metabolic syndrome, identification of intrinsic molecular programs responsible for ensuring fuel homeostasis and preventing metabolic disease is needed. We investigated whether the orphan nuclear receptor estrogen-related receptor α (ERRα), a major regulator
Ana Mato-Berciano et al.
Oncotarget, 8(14), 22700-22715 (2017-02-12)
Notch signaling pathway is an embryonic program that becomes reactivated in pancreatic cancer and contributes to cancer stem cell (CSC) maintenance. We explored the concept of oncolytic adenoviral activity in response to Notch activation signaling, in the context of a
M M Vick et al.
Journal of animal science, 85(5), 1144-1155 (2007-02-01)
Recent studies associate obesity and insulin resistance in horses with development of abnormal reproductive function and debilitating laminitis. The factors contributing to insulin resistance in obese horses are unknown. However, human studies provide evidence that elevated inflammatory cytokines such as
Martin Peterka et al.
PLoS genetics, 15(3), e1008050-e1008050 (2019-03-12)
CENP-F is a large, microtubule-binding protein that regulates multiple cellular processes including chromosome segregation and mitochondrial trafficking at cytokinesis. This multiplicity of functions is mediated through the binding of various partners, like Bub1 at the kinetochore and Miro at mitochondria.
Charlotte M François et al.
Scientific reports, 7, 46222-46222 (2017-04-12)
In cyclic females, FSH stimulates ovarian estradiol (E2) production and follicular growth up to the terminal stage. A transient elevation in circulating FSH and E2 levels occurs shortly after birth. But what could be the action of FSH on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service