Skip to Content
Merck
All Photos(4)

Key Documents

M5266

Sigma-Aldrich

Meglumine diatrizoate

organic

Synonym(s):

N-Methyl-D-glucamine diatrizoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17NO5 · C11H9I3N2O4
CAS Number:
Molecular Weight:
809.13
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

form

powder

optical activity

[α]25/D -6.4 to -5.7 °, c = 1% (w/v) in water

SMILES string

CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.CC(=O)Nc1c(I)c(NC(C)=O)c(I)c(C(O)=O)c1I

InChI

1S/C11H9I3N2O4.C7H17NO5/c1-3(17)15-9-6(12)5(11(19)20)7(13)10(8(9)14)16-4(2)18;1-8-2-4(10)6(12)7(13)5(11)3-9/h1-2H3,(H,15,17)(H,16,18)(H,19,20);4-13H,2-3H2,1H3/t;4-,5+,6+,7+/m.0/s1

InChI key

MIKKOBKEXMRYFQ-WZTVWXICSA-N

General description

Meglumine Diatrizoate is an organic, iodinated meglumine salt form of diatrizoate. It is also known as N-Methyl-D-glucamine diatrizoate.

Application

Meglumine diatrizoate can be used to treat long-term Taenia saginata infection. It has also been used to tag fecal matter and bodily fluids in a study investigating preoperative examination techniques in patients with acute colon obstruction caused by colorectal cancer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min A Yoon et al.
Journal of computer assisted tomography, 36(5), 596-601 (2012-09-21)
To evaluate the diagnostic performance of computed tomography (CT) colonography (CTC) reconstructed with different levels of adaptive statistical iterative reconstruction (ASiR, GE Healthcare) and Veo (model-based iterative reconstruction, GE Healthcare) at various tube currents in detection of polyps in porcine
A Balbir-Gurman et al.
Internal medicine journal, 42(3), 323-329 (2012-03-22)
Pneumatosis cystoides intestinalis (PCI) is a rare life-threatening gastrointestinal complication in the course of connective tissue disease (CTD). PCI is characterised by the appearance of intramural clusters of gas in the small and large bowel wall on X-ray or computed
A Pequignot et al.
Obesity surgery, 22(5), 712-720 (2012-02-14)
Laparoscopic sleeve gastrectomy (LSG) has a specific morbidity profile in which gastric leak (GL) is the main complication. With a view to defining a standardized protocol for GL management, the present retrospective study sought to describe the clinical patterns of
Zara M Patel et al.
International forum of allergy & rhinology, 2(4), 303-308 (2012-04-25)
Bacterial biofilms have been implicated in refractory rhinosinusitis. Biofilms have been shown to respond to treatment with low-frequency ultrasound (LFU) therapy in vitro, and exposure to LFU has shown efficacy in wound repair and topical drug delivery in other fields.
Nadja Haiden et al.
Pediatrics, 130(6), e1600-e1606 (2012-11-28)
To determine whether enteral application of the osmotic contrast agent Gastrografin accelerates complete meconium excretion and improves feeding tolerance in very low birth weight infants. This study was a stratified, randomized, placebo-controlled trial in premature infants with a birth weight

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service