Skip to Content
Merck
All Photos(6)

Key Documents

S4506

Sigma-Aldrich

Sodium diatrizoate hydrate

≥98.0% purity, powder

Synonym(s):

3,5-Bis(acetylamino)-2,4,6-triiodobenzoic acid, sodium salt, 3,5-Diacetamido-2,4,6-triiodobenzoic acid sodium salt, Diatrizoic acid sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8I3N2NaO4 · xH2O
CAS Number:
Molecular Weight:
635.90 (anhydrous basis)
Beilstein:
4082984
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

Product Name

Sodium diatrizoate hydrate, ≥98.0%

Assay

≥98.0%

form

powder

composition

Water (by Karl Fischer): ≤ 11.0%

technique(s)

titration: suitable

color

white to off-white

mp

261 °C

solubility

H2O: 350 mg/mL, clear to slightly hazy

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

O.[Na+].CC(=O)Nc1c(I)c(NC(C)=O)c(I)c(C([O-])=O)c1I

InChI

1S/C11H9I3N2O4.Na.H2O/c1-3(17)15-9-6(12)5(11(19)20)7(13)10(8(9)14)16-4(2)18;;/h1-2H3,(H,15,17)(H,16,18)(H,19,20);;1H2/q;+1;/p-1

InChI key

ZZBYVHJZBJEHFI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium diatrizoate hydrate has been used in spore purification. It has also been used in the isolation of plasma lymphocytes, monocytes, and dendritic cells.Sodium diatrizoate is also suitable as a density gradient reagent in blood cell separation. It has also been utilized as a radiological contrast agent.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Diffusion of MRI and CT contrast agents in articular cartilage under static compression.
Shafieyan Y, et al.
Biophysical Journal, 107(2), 485-492 (2014)
A Martinez et al.
Journal of chromatography. A, 1590, 65-72 (2019-01-15)
A major limitation of the three-dimensional imaging of polymeric biochromatography particle packings using X-ray computed tomography is that the particles have a low density and a high porosity, making them almost undistinguishable from the surrounding liquid phase. Additionally, the employed
Julia Jando et al.
PloS one, 12(9), e0184845-e0184845 (2017-09-16)
Absorption of neutral amino acids across the luminal membrane of intestinal enterocytes is mediated by the broad neutral amino acid transporter B0AT1 (SLC6A19). Its intestinal expression depends on co-expression of the membrane-anchored peptidase angiotensin converting enzyme 2 (ACE2) and is
Sandra Eickhoff et al.
Veterinary immunology and immunopathology, 101(1-2), 109-121 (2004-07-21)
Quantitative analysis of phagocytosis and oxidative burst in canine polymorphonuclear (PMN) cells was performed by flow cytometry techniques. Different concentrations of phorbol myristate acetate (PMA) were used to modulate PMN phagocytosis. A low concentration of PMA (3 nmol) resulted in
Y Luo et al.
Current protocols in immunology, Chapter 3, Unit 3-Unit 3 (2008-04-25)
Neutrophils are widely used in research to elucidate the mechanisms of inflammation, including such processes as cell adhesion, chemotaxis, superoxide release, production of reactive nitrogen intermediates, and granule exocytosis. This unit presents a protocol for the induction of an enriched

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service