Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C11H9I3N2O4
CAS Number:
Molecular Weight:
613.91
NACRES:
NA.75
PubChem Substance ID:
UNSPSC Code:
12352207
EC Number:
204-223-6
MDL number:
Product Name
Diatrizoic acid, Iodine-containing contrast agent
Quality Segment
form
powder
composition
Water (by Karl Fischer): ≤ 7.1%
concentration
≥98.0% AgNO3
technique(s)
titration: suitable
color
white to off-white
solubility
1 M NH4OH: 100 mg/mL, colorless
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
CC(=O)Nc1c(I)c(NC(C)=O)c(I)c(C(O)=O)c1I
InChI
1S/C11H9I3N2O4/c1-3(17)15-9-6(12)5(11(19)20)7(13)10(8(9)14)16-4(2)18/h1-2H3,(H,15,17)(H,16,18)(H,19,20)
InChI key
YVPYQUNUQOZFHG-UHFFFAOYSA-N
General description
Diatrizoic acid is an iodine-containing contrast agent. It is also known as amidotrizoic acid. Diatrizoic acid is one of the trace contaminants of waste water treatment plants.
Application
Diatrizoic acid has been used as a contrast-enhancing fluid for X-ray based measurements.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| D9268-50G | 04061833590553 |