Skip to Content
Merck
All Photos(2)

Documents

74147

Supelco

Iohexol

analytical standard

Synonym(s):

5-[N-(2,3-Dihydroxypropyl)acetamido]-2,4,6-triiodo-N,N′-bis(2,3-dihydroxypropyl)isophthalamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26I3N3O9
CAS Number:
Molecular Weight:
821.14
Beilstein:
2406632
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤6.0% water

application(s)

clinical testing

format

neat

SMILES string

IC1=C(C(NCC(O)CO)=O)C(I)=C(C(NCC(CO)O)=O)C(I)=C1N(CC(CO)O)C(C)=O

InChI

1S/C19H26I3N3O9/c1-8(29)25(4-11(32)7-28)17-15(21)12(18(33)23-2-9(30)5-26)14(20)13(16(17)22)19(34)24-3-10(31)6-27/h9-11,26-28,30-32H,2-7H2,1H3,(H,23,33)(H,24,34)

InChI key

NTHXOOBQLCIOLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iohexol is a non-ionic, water-soluble contrast agent used as a reference marker to determine the glomerular filtration rate (GFR) which measures the level of kidney function. It finds applications in angiographic and urography procedures.

Application

Iohexol may be used as an internal standard for the determination of iohexol in human plasma, serum and urine samples by HPLC-UV method. It may also be used as a model compound to analyze the applicability of iohexol for glomerular filtration rate (GFR) measurement by the plasma clearance of iohexol in investigating renal failure.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Performance of iohexol determination in serum and urine by HPLC: validation, risk and uncertainty assessment.
Cavalier E, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 396(1-2), 80-85 (2008)
Development and validation of an HPLC-UV method for determination of iohexol in human plasma.
Soman R S, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 816(1-2), 339-343 (2005)
Human pharmacokinetics of iohexol. A new nonionic contrast medium.
BO, et al.
Investigative Radiology, 18(2), 177-182 (1983)
Iohexol clearance for GFR-determination in renal failure?single or multiple plasma sampling?.
Sterner G, et al.
Nephrology, Dialysis, and Transplantation, 11(3), 521-525 (1996)
Claire Francoz et al.
Hepatology (Baltimore, Md.), 59(4), 1514-1521 (2013-09-17)
Simultaneous liver and kidney transplantation (SLKT) remains the procedure of choice for patients with both endstage liver disease and kidney failure. Stringent guidelines are needed to avoid unnecessary kidney transplantation. A recent consensus meeting proposed criteria based on the Modified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service