Skip to Content
Merck
All Photos(1)

Documents

3802

Sigma-Aldrich

Glutaraldehyde solution

for in vitro diagnostic use

Synonym(s):

Glutaric dialdehyde solution, Pentane-1,5-dial

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OHC(CH2)3CHO
CAS Number:
Molecular Weight:
100.12
Beilstein:
605390
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.28

form

solution

Quality Level

IVD

for in vitro diagnostic use

shipped in

wet ice

storage temp.

2-8°C

SMILES string

[H]C(CCCC([H])=O)=O

InChI

1S/C5H8O2/c6-4-2-1-3-5-7/h4-5H,1-3H2

InChI key

SXRSQZLOMIGNAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used as the fixative in the Sudan Black B staining procedure 380.

Other Notes

Glutaraldehyde, 4%, in borate buffer, pH 7.6.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Maertens et al.
BMC veterinary research, 16(1), 140-140 (2020-05-20)
Disinfectants are frequently used in animal production to reduce or eliminate the load of infectious agents and parasites in buildings and equipment associated with the housing or transportation of animals. There are growing concerns that the use of disinfectants would

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service