Skip to Content
Merck
All Photos(2)

Documents

L8629

Sigma-Aldrich

Lectin from Phaseolus vulgaris (red kidney bean)

Erythroagglutinin PHA-E

Synonym(s):

Phaseolus vulgaris agglutinin, PHA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

Quality Level

form

salt-free, lyophilized powder

potency

<10 μg per mL agglutination activity (with erythryocytes)

composition

Protein, ≥80% biuret

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

PHA consists of two molecular species, an erythroagglutinin (PHA-E) which has low mitogenic activity and high erythroagglutinin activity, and leucoagglutinin (PHA-L) which has high mitogenic and leucoagglutinating activity, but very low erythroagglutinating activity.
PHA-E is not blood group specific, but agglutination can be inhibited by certain oligosaccharides. PHA-P is the protein form of PHA prior to separation and purification of erythroagglutinin and leucoagglutinin. PHA-M is the mucoprotein form. Conjugates are prepared from the corresponding purified lectins.

Quality

Tested for leucoagglutination.

Preparation Note

Purified by a modification of the method of Leavitt.

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a suspension of either human erythrocytes (2% in phosphate buffered saline, pH 6.8) or human leukocytes (107 per mL in saline) after 1 hr incubation at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tuo Yang et al.
Cell reports, 29(9), 2917-2928 (2019-11-28)
Increased tolerance of Plasmodium falciparum to front-line artemisinin antimalarials (ARTs) is associated with mutations in Kelch13 (K13), although the precise role of K13 remains unclear. Here, we show that K13 mutations result in decreased expression of this protein, while mislocalization
R D Leavitt et al.
The Journal of biological chemistry, 252(9), 2961-2966 (1977-05-10)
Affinity-purified phytohemagglutinin from red kidney bean resolves into five isolectins by SP-Sephadex ion exchange chromatography. Recoveries ranging from 30 to 130 mg of protein for each isolectin are easily achieved. The isolectins have similar amino acid compositions which differ only
Malvina Prapa et al.
NPJ precision oncology, 5(1), 93-93 (2021-10-29)
Glioblastoma is the most malignant primary brain tumor and is still in need of effective medical treatment. We isolated patient-derived glioblastoma cells showing high GD2 antigen expression representing a potential target for CAR T strategy. Data highlighted a robust GD2
H J Hoffmann et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 33(7), 930-935 (2003-07-16)
Atopy is closely associated with the cellular T helper type-2 (Th2) phenotype, that is dominated by the pleiotrophic cytokine IL-4. The cellular source of IL-4 has yet to be determined, although basophils have been proposed. Eosinophils and mast cells are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service