Skip to Content
Merck
All Photos(1)

Key Documents

L1668

Sigma-Aldrich

Lectin from Phaseolus vulgaris (red kidney bean)

Phytohemagglutinin PHA-P, lyophilized powder (contains ~50% buffer salts as sodium phosphate and NaCl), BioReagent, suitable for cell culture

Synonym(s):

Phaseolus vulgaris agglutinin, PHA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

Quality Level

product line

BioReagent

form

lyophilized powder (contains ~50% buffer salts as sodium phosphate and NaCl)

potency

≤10 μg per mL mitogenic activity (Bromdeoxyuridine incorporation in human peripheral blood mononuclear cells)

technique(s)

cell culture | mammalian: suitable

color

white to off-white

solubility

PBS: 5 mg/mL, clear to slightly hazy, colorless to faintly yellow (pH 6.8)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Lectin from Phaseolus vulgaris (red kidney bean) has been used:
  • for the stimulation of cluster of differentiation (CD4+) T cells
  • for the activation of primary peripheral blood lymphocytes
  • for the stimulation of peripheral blood mononuclear cells

Biochem/physiol Actions

PHA consists of two molecular species, an erythroagglutinin (PHA-E) which has low mitogenic activity and high erythroagglutinin activity, and leucoagglutinin (PHA-L) which has high mitogenic and leucoagglutinating activity, but very low erythroagglutinating activity.
PHA-E is not blood group specific, but agglutination can be inhibited by certain oligosaccharides. PHA-P is the protein form of PHA prior to separation and purification of erythroagglutinin and leucoagglutinin. PHA-M is the mucoprotein form. Conjugates are prepared from the corresponding purified lectins.

Packaging

Sold on the basis of lectin content

Analysis Note

Potency and solubility expressed in mg lectin/ml

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nina Thakkar et al.
Antimicrobial agents and chemotherapy, 53(2), 631-638 (2008-12-03)
The present studies were conducted to better define the mechanism of action of polyethylene hexamethylene biguanide (PEHMB) (designated herein as NB325), which was shown in previous studies to inhibit infection by the human immunodeficiency virus type 1 (HIV-1). Fluorescence-activated flow
Cellular RelB interacts with the transactivator Tat and enhance HIV-1 expression
Wang M, et al.
Retrovirology, 15(1), 65-65 (2018)
Ema T Crooks et al.
PLoS pathogens, 14(5), e1007024-e1007024 (2018-05-03)
The extensive glycosylation of HIV-1 envelope (Env) glycoprotein leaves few glycan-free holes large enough to admit broadly neutralizing antibodies (bnAb). Consequently, most bnAbs must inevitably make some glycan contacts and avoid clashes with others. To investigate how Env glycan maturation
Primate immunodeficiency virus proteins Vpx and Vpr counteract transcriptional repression of proviruses by the HUSH complex
Yurkovetskiy L, et al.
Nature microbiology, 1-1 (2018)
Posttranscriptional Regulation of HIV-1 Gene Expression during Replication and Reactivation from Latency by Nuclear Matrix Protein MATR3
Sarracino A, et al.
mBio, 9(6), e02158-e02118 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service