Skip to Content
Merck
All Photos(2)

Key Documents

L8902

Sigma-Aldrich

Lectin from Phaseolus vulgaris (red kidney bean)

Phytohemagglutinin PHA-M, lyophilized powder, BioReagent, suitable for cell culture

Synonym(s):

Phaseolus vulgaris agglutinin, PHA

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352202
NACRES:
NA.32

Quality Level

product line

BioReagent

form

lyophilized powder

potency

<40 μg per mL agglutination activity

technique(s)

cell culture | mammalian: suitable

impurities

salt, free

solubility

PBS: 2 mg/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Lectin from Phaseolus vulgaris (red kidney bean) is used in Lymphocyte Transformation Assay and culture medium.

Biochem/physiol Actions

PHA consists of two molecular species, an erythroagglutinin (PHA-E) which has low mitogenic activity and high erythroagglutinin activity, and leucoagglutinin (PHA-L) which has high mitogenic and leucoagglutinating activity, but very low erythroagglutinating activity.
PHA-E is not blood group specific, but agglutination can be inhibited by certain oligosaccharides. PHA-P is the protein form of PHA prior to separation and purification of erythroagglutinin and leucoagglutinin. PHA-M is the mucoprotein form. Conjugates are prepared from the corresponding purified lectins.

Analysis Note

Agglutination activity is expressed in μg/mL and is determined from serial dilutions of a 1 mg/mL solution. This activity is the lowest concentration to agglutinate a 2% suspension of human erythrocytes in phosphate buffered saline, pH 6.8, after 1 hr incubation at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan Warmuth et al.
Oncoimmunology, 10(1), 2004661-2004661 (2022-07-19)
Co-stimulatory 4-1BB receptors on tumor-infiltrating T cells are a compelling target for overcoming resistance to immune checkpoint inhibitors, but initial clinical studies of 4-1BB agonist mAbs were accompanied by liver toxicity. We sought to engineer a tri-specific antibody-based molecule that
Nina Li et al.
iScience, 24(10), 103163-103163 (2021-10-26)
RANTES is implicated in allergic asthma and in T cell-dependent clearance of infection. RANTES receptor family comprises CCR1, CCR3, and CCR5, which are G-protein-coupled receptors consisting of seven transmembrane helices. Infections with respiratory viruses like Rhinovirus cause induction of RANTES production by
Imen Helali et al.
Journal of immunotoxicology, 13(5), 745-750 (2016-07-19)
The immune response is the first defense against pathogens; however, it is very sensitive and can be impacted on by agrochemicals such as carbamate and organophosphate pesticides widely present in the environment. To understand how pesticides can affect immune cell
Packaging cell system for lentivirus vectors. Preparation and use.
Narasimhachar Srinivasakumar
Methods in molecular medicine, 69, 275-302 (2002-05-04)
Sarah Chan et al.
Nature cancer, 3(3), 337-354 (2022-03-09)
Costimulatory receptors such as glucocorticoid-induced tumor necrosis factor receptor-related protein (GITR) play key roles in regulating the effector functions of T cells. In human clinical trials, however, GITR agonist antibodies have shown limited therapeutic effect, which may be due to suboptimal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service