Skip to Content
Merck
All Photos(1)

Key Documents

11082132001

Roche

Phytohemagglutinin-M (PHA-M)

from Phaseolus vulgaris

Synonym(s):

PHA-M

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
Pricing and availability is not currently available.

Quality Level

sterility

non-sterile; 0.2 μm filtered

form

lyophilized

packaging

pkg of 20 mg

manufacturer/tradename

Roche

application(s)

hematology

storage temp.

2-8°C

Related Categories

General description

Phytohemagglutinin (PHA), the lectin extract from the red kidney bean. PHA consists of five isolectins (L4E0, L3E1, L2E2, L1E3, L0E4) each being a tetramer held together by noncovalent forces. The subunits L (leukocyte reactive) have a high affinity for lymphocyte surface receptors, but little for those of erythrocytes, and are responsible for the mitogenic properties of the isolectins. E (erythrocyte reactive) is responsible for the erythrocyte-agglutinating properties.[1]

Application

Phytohemagglutinin (PHA) is a potent mitogen used to stimulate cell proliferation in lymphocyte cultures.[2]
Phytohemagglutinin-M (PHA-M) has been used in the activation of Jurkat and primary cells.[3] It has also been used to treat polymorphonuclear blood cells(PMBCs)[4]

Biochem/physiol Actions

Phytohemagglutinin (PHA) has mitogenic and potent cell agglutinating activities. It binds to T-cell membranes and stimulates cell division and metabolic activity. PHA has the ability to stimulate close contacts between cell membranes.[1]

Specifications

Biological activity: <10 μg/ml for maximal stimulation of DNA synthesis (BrdU incorporation) with human peripheral blood lymphocytes.

Preparation Note

Working concentration: 2 - 10 μg/ml
Approximately 2 - 10 μg/ml for the stimulation of peripheral blood lymphocytes
Storage conditions (working solution): The reconstituted solution is stable for 2 weeks at 2 to 8 °C or for several months when stored in aliquots at -15 to -25 °C.

Reconstitution

PHA-M should be reconstituted in sterile double-dist. water (final concentration: 2–10 mg/ml). Further dilution with medium or PBS (phosphate buffered saline).

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia Kölle et al.
iScience, 25(6), 104440-104440 (2022-06-17)
The cytokine interleukin-3 (IL-3) acts on early hematopoietic precursor cells. In humans, Treg cells secrete IL-3 and repress inflammatory cells except for basophils. The present study aims to elucidate the contribution of IL-3 in the development and the course of
Gilles Darcis et al.
Cell reports, 30(7), 2284-2296 (2020-02-23)
The HIV latent reservoir forms the major hurdle to an HIV cure. The discovery of CD32 as marker of this reservoir has aroused much interest, but subsequent reports have challenged this finding. Here, we observe a positive correlation between the
Polyfunctional Melan-A-specific tumor-reactive CD8+ T cells elicited by dacarbazine treatment before peptide-vaccination depends on AKT activation sustained by ICOS
Ornella Franzese
Oncoimmunology, 5(5), e1114203-e1114203 (2016)
TRIM28 promotes HIV-1 latency by SUMOylating CDK9 and inhibiting P-TEFb
Ma X, et al.
eLife, 8, e42426-e42426 (2019)
Tania Del Rivero et al.
Frontiers in immunology, 13, 977809-977809 (2022-12-16)
Extracellular vesicles isolated from human amniotic fluid (AF-EVs) have previously been found to modulate inflammation and macrophage infiltration in a mouse model. However, the effects of acellular amniotic fluid (acAF) or AF-EVs on the T-Cell immune response have not been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service