L9640
Lectin from Triticum vulgaris (wheat)
lyophilized powder
Synonym(s):
WGA, Wheat germ agglutinin
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
form
lyophilized powder
Quality Level
potency
<20 μg/mL agglutination activity
composition
Protein, 100% biuret
impurities
salt, essentially free
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
Biochem/physiol Actions
WGA is not blood group specific but has an affinity for N-acetyl-β-D-glucosaminyl residues and N-acetyl-β-D-glucosamine oligomers. WGA contains no protein-bound carbohydrate.
Preparation Note
Conjugates are prepared from affinity purified lectin.
Analysis Note
Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a 2% suspension of human erythrocytes after 1 hour incubation at 25 °C.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jiamao Lin et al.
European journal of pharmacology, 859, 172490-172490 (2019-06-24)
Doxorubicin (DOX) is a classic anti-tumor chemotherapeutic used to treat a wide range of tumors. One major downfall of DOX treatment is it can induce fatal cardiotoxicity. Astragaloside IV (AS-IV) is one of the primary active ingredients that can be
André A Neves et al.
Endoscopy, 50(6), 618-625 (2018-01-18)
Endoscopic surveillance for Barrett's esophagus (BE) is limited by long procedure times and sampling error. Near-infrared (NIR) fluorescence imaging minimizes tissue autofluorescence and optical scattering. We assessed the feasibility of a topically applied NIR dye-labeled lectin for the detection of
Qian Li et al.
Foods (Basel, Switzerland), 10(12) (2021-12-25)
Paeonol can effectively inhibit Aspergillus flavus (A. flavus) via damaging cell walls. In this work, paeonol treatment remarkably destroyed both the outer amorphous layer and the inner fibrous layer of cell walls. Furthermore, FT-IR and XPS characterization showed that OH
Ilaria Volpi et al.
Open biology, 11(10), 210121-210121 (2021-10-27)
Treslin/Ticrr is required for the initiation of DNA replication and binds to MTBP (Mdm2 Binding Protein). Here, we show that in Xenopus egg extract, MTBP forms an elongated tetramer with Treslin containing two molecules of each protein. Immunodepletion and add-back
Julie Beneteau et al.
Plant physiology, 153(3), 1345-1361 (2010-05-06)
Phloem Protein2 (PP2) is a component of the phloem protein bodies found in sieve elements. We describe here the lectin properties of the Arabidopsis (Arabidopsis thaliana) PP2-A1. Using a recombinant protein produced in Escherichia coli, we demonstrated binding to N-acetylglucosamine
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service