Skip to Content
Merck
All Photos(2)

Key Documents

A9518

Sigma-Aldrich

Ampicillin sodium salt

Synonym(s):

D-(−)-α-Aminobenzylpenicillin sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N3NaO4S
CAS Number:
Molecular Weight:
371.39
Beilstein:
4119211
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.76

biological source

synthetic (chemical)

Quality Level

form

powder

color

white to off-white

mp

215 °C (dec.) (lit.)

solubility

water: 50 g/L

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

agriculture

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[Na+].CC1(C)SC2[C@H](NC(=O)[C@H](N)c3ccccc3)C(=O)N2[C@H]1C([O-])=O

InChI

1S/C16H19N3O4S.Na/c1-16(2)11(15(22)23)19-13(21)10(14(19)24-16)18-12(20)9(17)8-6-4-3-5-7-8;/h3-7,9-11,14H,17H2,1-2H3,(H,18,20)(H,22,23);/q;+1/p-1/t9-,10-,11+,14-;/m1./s1

InChI key

KLOHDWPABZXLGI-YWUHCJSESA-M

Looking for similar products? Visit Product Comparison Guide

General description

Ampicillin sodium salt is a member of the extended-spectrum β-lactam family and a semisynthetic derivative of penicillin that functions as a broad-spectrum antibiotic. It is a β-lactam antibiotic that inhibits bacterial cell-wall synthesis by binding to penicillin-binding proteins (PBPs), thereby inhibiting peptidoglycan synthesis, a critical component of the bacterial cell wall. Ampicillin sodium salt is active against a wide range of Gram-positive and Gram-negative bacteria, including E. coli, β-lactam-sensitive VRE, vancomycin-resistant Enterococcus species, Staphylococcus aureus, and Streptococcus pneumoniae. It is commonly used in research laboratories to study antibiotic resistance and penetration limitations, the synergy between multiple antibiotics, and the selection and maintenance of recombinant plasmids in E. coli.

Application

Ampicillin sodium salt has been used:
  • to select for ampicillin resistance in mutated and transformed cells

Biochem/physiol Actions

Mode of Action: This is a ß-lactam antibiotic that inhibits bacterial cell-wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane.

Mode of Resistance: Administration with ß-lactamase cleaves the ß-lactam ring of Ampicillin and inactivates it.

Antimicrobial Spectrum: Includes both gram-positive (similar to benzylpenicillin) and gram-negative bacteria (similar to tetracyclines and chloramphenicol.

Features and Benefits

  • High-quality antibiotic suitable for multiple research applications
  • Ideal for Cell Biology and Biochemical research.

Caution

This product has been reported stable as supplied at 25°C at 43% and 81% relative humidity for six weeks. Additional studies have shown that the stability of Ampicillin in solution is a function of pH, temperature and the identity of the buffer. It′s activity is quickly lost when stored above pH 7. Optimal storage conditions are suggested as 2-8°C, and pH 3.8-5 where its activity was retained at 90%+ for a week.

Preparation Note

Ampicillin is reported as slightly soluble in water, practically insoluble in alcohol, chloroform, ether and fixed oils but soluble in dilute acids or bases. The solution should not be autoclaved; a stock solution should be sterilized through filtration and stored frozen, where it will be stable for months.

Storage and Stability

Tightly closed. Dry. Keep locked up or in an area accessible only to qualified or authorized

Other Notes

For additional information on our range of Biochemicals, please complete this form.
Keep container tightly closed in a dry and well-ventilated place. Storage class (TRGS 510): Non Combustible Solids

comparable product

Product No.
Description
Pricing

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1A - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christoffer V Sørensen et al.
Toxins, 12(4) (2020-04-15)
Antibiotics are often administered with antivenom following snakebite envenomings in order to avoid secondary bacterial infections. However, to this date, no studies have evaluated whether antibiotics may have undesirable potentiating effects on snake venom. Herein, we demonstrate that four commonly
Nuria Fernández-Hidalgo et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 56(9), 1261-1268 (2013-02-09)
The aim of this study was to compare the effectiveness of the ampicillin plus ceftriaxone (AC) and ampicillin plus gentamicin (AG) combinations for treating Enterococcus faecalis infective endocarditis (EFIE). An observational, nonrandomized, comparative multicenter cohort study was conducted at 17
Nikos Fatsis-Kavalopoulos et al.
PLoS biology, 18(9), e3000856-e3000856 (2020-09-18)
Antibiotic combination therapies are important for the efficient treatment of many types of infections, including those caused by antibiotic-resistant pathogens. Combination treatment strategies are typically used under the assumption that synergies are conserved across species and strains, even though recent
Bram Lories et al.
Current biology : CB, 30(7), 1231-1244 (2020-02-23)
Bacteria use complex regulatory networks to cope with stress, but the function of these networks in natural habitats is poorly understood. The competition sensing hypothesis states that bacterial stress response systems can serve to detect ecological competition, but studying regulatory
Ritesh Ranjan Pal et al.
Cell, 177(3), 683-696 (2019-04-02)
Microbiota and intestinal epithelium restrict pathogen growth by rapid nutrient consumption. We investigated how pathogens circumvent this obstacle to colonize the host. Utilizing enteropathogenic E. coli (EPEC), we show that host-attached bacteria obtain nutrients from infected host cell in a process we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service