Skip to Content
Merck
All Photos(2)

Documents

L2884

Sigma-Aldrich

Leupeptin

powder, ≥90% (HPLC)

Synonym(s):

Leupeptin hemisulfate salt, Acetyl-Leu-Leu-Arg-al, N-Acetyl-L-leucyl-L-leucyl-L-argininal hemisulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C20H38N6O4 · 1/2H2SO4
CAS Number:
Molecular Weight:
475.59
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

product name

Leupeptin, microbial, ≥90% (HPLC)

biological source

microbial

Quality Level

Assay

≥90% (HPLC)

form

powder

solubility

H2O: 10 mM (Solutions are stable for a week at 4 °C. Stock solutions are stable up to 6 months at −20 °C.)
H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

OS(O)(=O)=O.CC(C)C[C@H](NC(C)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C=O.CC(C)C[C@H](NC(C)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C=O

InChI

1S/2C20H38N6O4.H2O4S/c2*1-12(2)9-16(24-14(5)28)19(30)26-17(10-13(3)4)18(29)25-15(11-27)7-6-8-23-20(21)22;1-5(2,3)4/h2*11-13,15-17H,6-10H2,1-5H3,(H,24,28)(H,25,29)(H,26,30)(H4,21,22,23);(H2,1,2,3,4)/t2*15-,16-,17-;/m00./s1

InChI key

CIPMKIHUGVGQTG-VFFZMTJFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Leupeptin, or N-acetyl-L-leucyl-L-leucyl-L-argininal, is a naturally occurring tripeptide that acts particularly as a serine protease inhibitor and as a cysteine protease inhibitor. Its mechanism of action involves structurally similar covalent binding reactions:
  • In the active site of serine proteases, leupeptin forms a covalent hemiacetal adduct between the aldehyde group of leupeptin and the hydroxyl group of a serine residue in the enzyme active site.
  • In the active site of cysteine proteases, the electrophilic (aldehyde) carbon of leupeptin forms a comparable bond with the sulfur atom of a cysteine residue in the enzyme active site.

Biochem/physiol Actions

Inhibitor of serine and cysteine proteases. Inhibits plasmin, trypsin, papain, calpain, and cathepsin B. Does not inhibit pepsin, cathepsins A and D, thrombin, or α-chymotrypsin. Effective concentration 10-100 μM. There have been numerous studies using leupeptin to protect against hearing loss caused by acoustic overstimulation or the ototoxic antibiotic gentamicin. (Loss of cochlear hair cells is believed to be mediated by calpain.)

Analysis Note

Leupeptin gives multiple peaks on HPLC because of the formation of tautomeric isomers in solution. Majority of contaminating peptide is racemized leupeptin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shohreh Majd et al.
BMC cell biology, 19(1), 7-7 (2018-06-21)
Cellular energy failure in high metabolic rate organs is one of the underlying causes for many disorders such as neurodegenerative diseases, cardiomyopathies, liver and renal failures. In the past decade, numerous studies have discovered the cellular axis of LKB1/AMPK/mTOR as
Yang Yang et al.
Journal of dairy science, 100(5), 4014-4024 (2017-03-13)
Adequate lipid synthesis by the mammary gland during lactation is essential for the survival of mammalian offspring. Cell death-inducing DNA fragmentation factor-α-like effector C (CIDEC) is a lipid droplet-associated protein and functions to promote lipid accumulation and inhibit lipolysis in
Satoshi Nakamura et al.
Scientific reports, 10(1), 12242-12242 (2020-07-24)
Vitamin D deficiency is a recognized risk factor for sarcopenia development, but mechanisms underlying this outcome are unclear. Here, we show that low vitamin D status worsens immobilization-induced muscle atrophy in mice. Mice globally lacking vitamin D receptor (VDR) exhibited
Wibke Schwarzer et al.
Human molecular genetics, 18(21), 4013-4021 (2009-07-31)
Mutations in ROR2 cause dominant brachydactyly type B (BDB1) or recessive Robinow syndrome (RRS), each characterized by a distinct combination of phenotypic features. We here report a novel nonsense mutation in ROR2 (c.1324C>T; p.R441X) causing intracellular protein truncation in a
Philipp A Jaeger et al.
Molecular cell, 69(2), 321-333 (2018-01-21)
We have developed a highly parallel strategy, systematic gene-to-phenotype arrays (SGPAs), to comprehensively map the genetic landscape driving molecular phenotypes of interest. By this approach, a complete yeast genetic mutant array is crossed with fluorescent reporters and imaged on membranes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service