Skip to Content
Merck
All Photos(2)

Documents

B8385

Sigma-Aldrich

Bestatin hydrochloride

powder, ≥98% (HPLC)

Synonym(s):

N-[(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyryl]-L-leucine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24N2O4 · HCl
CAS Number:
Molecular Weight:
344.83
Beilstein:
4628066
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

product name

Bestatin hydrochloride, ≥98% (HPLC)

biological source

synthetic (organic)

Assay

≥98% (HPLC)

form

powder

mp

216 °C

solubility

water: 25 mg/mL, clear to very slightly hazy, colorless

antibiotic activity spectrum

neoplastics

Mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

O=C(N[C@@H](CC(C)C)C(O)=O)[C@@H](O)[C@H](N)CC1=CC=CC=C1.Cl[H]

InChI

1S/C16H24N2O4.ClH/c1-10(2)8-13(16(21)22)18-15(20)14(19)12(17)9-11-6-4-3-5-7-11;/h3-7,10,12-14,19H,8-9,17H2,1-2H3,(H,18,20)(H,21,22);1H/t12-,13+,14+;/m1./s1

InChI key

XGDFITZJGKUSDK-UDYGKFQRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: amino acid derivatives

Biochem/physiol Actions

A metalloprotease inhibitor selective for aminopeptidase. Bestatin is a competitive and specific inhibitor of leucine aminopeptidase, aminopeptidase B, and triamino peptidase. It inhibits aminopeptidase B at 60 nM (using arginine-β-naphthylamide as substrate) and leucine aminopeptidase at 20 nM (leucine-β-naphthylamide as substrate). It showed no inhibition of aminopeptidase A, trypsin, chymotrypsin, elastase, papain, pepsin, or themolysin. It offers promise as a novel analgesic because it protects endogenous opioid peptides against degradation.

Preparation Note

Solubility testing in water at 25 mg/ml yields a clear solution. Stock solutions at 1 mM are expected to be stable at least one month stored at -20 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye Cheong Koo et al.
International journal of molecular sciences, 22(8) (2021-05-01)
It has been shown previously that a novel tetrapeptide, Arg-Leu-Tyr-Glu (RLYE), derived from human plasminogen inhibits vascular endothelial growth factor (VEGF)-induced angiogenesis, suppresses choroidal neovascularization in mice by an inhibition of VEGF receptor-2 (VEGFR-2) specific signaling pathway. In this study
A Taylor
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 7(2), 290-298 (1993-02-01)
Aminopeptidases catalyze the cleavage of amino acids from the amino terminus of protein or peptide substrates. They are widely distributed throughout the animal and plant kingdoms and are found in many subcellular organelles, in cytoplasm, and as membrane components. Several
Tina S Skinner-Adams et al.
Journal of medicinal chemistry, 50(24), 6024-6031 (2007-10-27)
Previous studies have pinpointed the M17 leucyl aminopeptidase of Plasmodium falciparum (PfLAP) as a target for the development of new antimalarials. This metallo-exopeptidase functions in the terminal stages of hemoglobin digestion and is inhibited by bestatin, a natural analog of
Anna L Epp et al.
Scientific reports, 9(1), 9721-9721 (2019-07-07)
The Pannexin 1 (Panx1) ion and metabolite channel is expressed in a wide variety of cells where it regulates a number of cell behaviours including proliferation and differentiation. Panx1 is expressed on the cell surface as well as intracellular membranes.
Yekaterina Poloz et al.
Eukaryotic cell, 11(4), 545-557 (2012-02-22)
Bestatin methyl ester (BME) is an inhibitor of Zn(2+)-binding aminopeptidases that inhibits cell proliferation and induces apoptosis in normal and cancer cells. We have used Dictyostelium as a model organism to study the effects of BME. Only two Zn(2+)-binding aminopeptidases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service