Merck
  • Vitamin D protects against immobilization-induced muscle atrophy via neural crest-derived cells in mice.
Vitamin D protects against immobilization-induced muscle atrophy via neural crest-derived cells in mice.
Scientific reports (2020-07-24)
Satoshi Nakamura, Yuiko Sato, Tami Kobayashi, Yosuke Kaneko, Eri Ito, Tomoya Soma, Hiroyuki Okada, Kana Miyamoto, Akihito Oya, Morio Matsumoto, Masaya Nakamura, Arihiko Kanaji, Takeshi Miyamoto
ABSTRACT
Vitamin D deficiency is a recognized risk factor for sarcopenia development, but mechanisms underlying this outcome are unclear. Here, we show that low vitamin D status worsens immobilization-induced muscle atrophy in mice. Mice globally lacking vitamin D receptor (VDR) exhibited more severe muscle atrophy following limb immobilization than controls. Moreover, immobilization-induced muscle atrophy was worse in neural crest-specific than in skeletal muscle-specific VDR-deficient mice. Tnfα expression was significantly higher in immobilized muscle of VDR-deficient relative to control mice, and was significantly elevated in neural crest-specific but not muscle-specific VDR-deficient mice. Furthermore, muscle atrophy induced by limb immobilization in low vitamin D mice was significantly inhibited in Tnfα-deficient mice. We conclude that vitamin D antagonizes immobilization-induced muscle atrophy via VDR expressed in neural crest-derived cells.
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Pepstatin A, microbial, ≥90% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid
Pricing and availability is not currently available.