L1882
Lectin from Triticum vulgaris (wheat)
agarose conjugate, saline suspension
Synonym(s):
WGA, Wheat germ agglutinin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
conjugate
agarose conjugate
Quality Level
form
saline suspension
extent of labeling
5-10 mg per mL
matrix
cross-linked 4% beaded agarose
matrix activation
cyanogen bromide
capacity
≥0.1 mg/mL binding capacity (ovomucoid)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Lectins are carbohydrate-binding proteins, found in fungi, plants and animals. The secondary structure of a lectin is rich in β-strands and possesses a carbohydrate binding sites on the surface.
Application
Used for the purification of glycoproteins containing (D-N-acetylglucosamine)2 or N-acetylneuraminic acid.
Biochem/physiol Actions
Lectins are known to be useful in glycoconjugate characterizing, imaging and targeting. They specifically interact with oligosaccharides, glycoproteins and glycolipids. Hence, lectins are used in microarray assays, enabling efficient glycome profiling. In plants and fungi, lectins defend against pathogens or feeders. They are also involved in host recognition and tissue adhesion.
WGA is not blood group specific but has an affinity for N-acetyl-β-D-glucosaminyl residues and N-acetyl-β-D-glucosamine oligomers. WGA contains no protein-bound carbohydrate.
Physical form
Suspension in 1.0 M NaCl containing 15 mM sodium azide
Preparation Note
Conjugates are prepared from affinity purified lectin.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
K Paz et al.
The Journal of biological chemistry, 271(12), 6998-7003 (1996-03-22)
A structural analysis has been carried out to determine which part of the intracellular domain of the insulin receptor (IR) beta subunit is involved in direct interaction with the receptor substrates IRS-1 and Shc. Toward this end, the juxtamembrane (JM)
Shohei Wakao et al.
Cellular and molecular life sciences : CMLS, 79(11), 542-542 (2022-10-07)
Stem cells undergo cytokine-driven differentiation, but this process often takes longer than several weeks to complete. A novel mechanism for somatic stem cell differentiation via phagocytosing 'model cells' (apoptotic differentiated cells) was found to require only a short time frame.
Binding sugars: from natural lectins to synthetic receptors and engineered neolectins
Arnaud J, et al.
Chemical Society Reviews, 42(11), 4798-4813 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service