L1394
Lectin from Triticum vulgaris (wheat)
agarose conjugate, saline suspension
Synonym(s):
WGA, Wheat germ agglutinin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
conjugate
agarose conjugate
Quality Level
form
saline suspension
extent of labeling
~6 mg per mL
matrix
4% agarose
matrix activation
cyanogen bromide
matrix spacer
1 atom
particle size
45-165 μm
capacity
≥0.5 mg/mL binding capacity (ovomucoid)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Used for the purification of glycoproteins containing (D-N-acetylglucosamine)2 or N-acetylneuraminic acid.
Biochem/physiol Actions
WGA is not blood group specific but has an affinity for N-acetyl-β-D-glucosaminyl residues and N-acetyl-β-D-glucosamine oligomers. WGA contains no protein-bound carbohydrate.
Physical form
Suspension in 0.9% NaCl containing 15 mM Sodium azide.
Preparation Note
Conjugates are prepared from affinity purified lectin.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
PloS one, 7(5), e34274-e34274 (2012-05-17)
There is controversy with respect to molecular characteristics of insulin analogues. We report a series of experiments forming a comprehensive characterisation of the long acting insulin analogues, glargine and detemir, in comparison with human insulin, IGF-1, and the super-mitogenic insulin
The Journal of biological chemistry, 291(8), 3905-3917 (2015-12-20)
The normal cellular prion protein (PrP) is a glycosylphosphatidylinositol (GPI)-anchored cell surface glycoprotein. However, in pancreatic ductal adenocarcinoma cell lines, such as BxPC-3, PrP exists as a pro-PrP retaining its glycosylphosphatidylinositol (GPI) peptide signaling sequence. Here, we report the identification
The Biochemical journal, 270(1), 57-61 (1990-08-15)
Chromaffin-granule membranes were separated into insoluble and soluble fractions after extraction with lithium di-iodosalicylate (LDIS). These fractions were characterized by one- and two-dimensional gel electrophoresis, and glycoproteins were detected after electroblotting with peroxidase-labelled concanavalin A and wheat-germ agglutinin (WGA). The
International journal of molecular sciences, 24(2) (2023-01-22)
In Alzheimer's disease (AD), the reduction in acetylcholinesterase (AChE) enzymatic activity is not paralleled with changes in its protein levels, suggesting the presence of a considerable enzymatically inactive pool in the brain. In the present study, we validated previous findings
Communications biology, 4(1), 161-161 (2021-02-07)
Mutations in CLN3 lead to photoreceptor cell loss in CLN3 disease, a lysosomal storage disorder characterized by childhood-onset vision loss, neurological impairment, and premature death. However, how CLN3 mutations cause photoreceptor cell death is not known. Here, we show that
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service