Skip to Content
Merck
All Photos(1)

Key Documents

L5142

Sigma-Aldrich

Lectin from Triticum vulgaris (wheat)

biotin conjugate, lyophilized powder

Synonym(s):

WGA, Wheat germ agglutinin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
34058011
NACRES:
NA.32

conjugate

biotin conjugate

Quality Level

form

lyophilized powder

composition

Protein, ≥80% E1%/280

extent of labeling

2-4 mol biotin per mol protein

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Wheat germ lectin (WGA) from Triticum vulgaris exists as a homodimer and has multiple N-acetyl glucosamine (GlcNAc) binding sites. It is resistant to protein denaturation agents and is used for the detection of O-GlcNAc proteins. WGA binding to N-glycolylneuraminic acid (Neu5Ac) may modulate pro-inflammatory immune response.

Application

Lectin from Triticum vulgaris (wheat) has been used:
  • in lectin binding assay of samples from intrauterine stages of filarial parasites
  • in the histological studies of mice myocytes
  • for the generation bead-biotin-WGA complex with Dynabeads® M-280 Streptavidin magnetic beads for lectin magnetic separation of Toxoplasma gondii oocytes from water samples

Biochem/physiol Actions

WGA is not blood group specific but has an affinity for N-acetyl-β-D-glucosaminyl residues and N-acetyl-β-D-glucosamine oligomers. WGA contains no protein-bound carbohydrate.

Packaging

Package size based on protein content.

Physical form

Contains sodium citrate

Preparation Note

Conjugates are prepared from affinity purified lectin.

Legal Information

Dynabeads is a registered trademark of Invitrogen Dynal AS Corp

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A flow cytometry based method for studying embryogenesis and immune reactivity to embryogenic stages in filarial parasites
Sahu BR, et al.
Filaria Journal, 4(1), 11-11 (2005)
The dietary intake of wheat and other cereal grains and their role in inflammation
De PK and Pruimboom L
Nutrients, 5(3), 771-787 (2013)
Evaluation of bi-ventricular coronary flow patterns using high-frequency ultrasound in mice with transverse aortic constriction
Wu J, et al.
Ultrasound in Medicine & Biology, 39(11), 2053-2065 (2013)
Candida Fagundes Teixeira et al.
PLoS neglected tropical diseases, 1(2), e73-e73 (2007-12-07)
Diagnosis of intestinal schistosomiasis in low endemic areas is a problem because often control measures have reduced egg burdens in feces to below the detection limits of classical coproparasitological methods. Evaluation of molecular methods is hindered by the absence of
R E Morris et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 40(5), 711-721 (1992-05-01)
We demonstrate here that the intracellular routing of biotinylated ligands was not affected by the attachment of streptavidin gold colloids so long as the electron-dense marker was added after the biotinyl ligand-receptor interaction had occurred. The binding, internalization, and intracellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service