L4895
Lectin from Triticum vulgaris (wheat)
FITC conjugate, lyophilized powder
Synonym(s):
WGA, Wheat germ agglutinin
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Recommended Products
conjugate
FITC conjugate
Quality Level
form
lyophilized powder
potency
<50 μg/mL agglutination activity
composition
Protein, ~50% Lowry
extent of labeling
~2 mol FITC per mol protein
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Lectins are carbohydrate-binding proteins. They are omnipresent and usually found in fungi, plants and animals. The secondary structure of this protein is rich in β-strands and possesses carbohydrate binding sites on the surface.
Application
For the fluorescent detection of glycoproteins containing β(1→4)-N-acetyl-D-glucosamine
Lectin from Triticum vulgaris (wheat) has been used:
- to treat transfected cells for immunofluorescence
- to stereotaxically inject into mice for neuronal tracing
- in immunohistochemistry for the analysis of myocytes
Biochem/physiol Actions
Lectin plays a key role in glycoconjugate characterizing, imaging and targeting. Its use in a microarray assay, enable efficient glycome profiling. In plants and fungi, lectin defends against pathogens/feeders. Lectin also plays a major role in host recognition and tissue adhesion.
WGA is not blood group specific but has an affinity for N-acetyl-β-D-glucosaminyl residues and N-acetyl-β-D-glucosamine oligomers. WGA contains no protein-bound carbohydrate.
Packaging
Package size based on protein content.
Physical form
Contains phosphate buffer salts
Preparation Note
Conjugates are prepared from affinity purified lectin.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
PloS one, 13(8), e0201986-e0201986 (2018-08-14)
Tissues with high-energy demand including the heart are rich in the energy-producing organelles, mitochondria, and sensitive to mitochondrial dysfunction. While alterations in mitochondrial function are increasingly recognized in cardiovascular diseases, the molecular mechanisms through which changes in mitochondria lead to
Science signaling, 12(577) (2019-04-18)
Stress is increasingly associated with heart dysfunction and is linked to higher mortality rates in patients with cardiometabolic disease. Glucocorticoids are primary stress hormones that regulate homeostasis through two nuclear receptors, the glucocorticoid receptor (GR) and mineralocorticoid receptor (MR), both
Novel Chemical Suppressors of Long QT Syndrome Identified by an In Vivo Functional Screen
Circulation (2011)
Scientific reports, 7(1), 8703-8703 (2017-08-20)
Diabetes is a major risk factor for cardiovascular disease and the lysosomal cysteine protease cathepsin K plays a critical role in cardiac pathophysiology. To expand upon our previous findings, we tested the hypothesis that, knockout of cathepsin K protects against
Langmuir : the ACS journal of surfaces and colloids, 26(8), 5603-5609 (2009-12-02)
The technique to pattern aminosilanes on hydroxyl-terminated substrates will open up extensive applications in many fields. There are some existing methods to pattern aminosilanes, in particular, (3-aminopropyl)triethoxysilane (APTES) on SiO(2) and glass substrates through indirect routes. However, few reports focus
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service