Skip to Content
Merck
All Photos(2)

Key Documents

H2270

Sigma-Aldrich

Hydrocortisone 21-hemisuccinate sodium salt

powder, BioReagent, suitable for cell culture

Synonym(s):

11β,17α,21-Trihydroxy-4-pregnene-3,20-dione 21-hemisuccinate sodium salt, Cortisol 21-hemisuccinate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H33NaO8
CAS Number:
Molecular Weight:
484.51
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

product line

BioReagent

Assay

≥90% (HPLC)

form

powder

technique(s)

cell culture | mammalian: suitable

solubility

H2O: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C1CC[C@@]2(C)C(CC[C@]3([H])[C@]2([H])[C@@H](O)C[C@@]4(C)[C@@]3([H])CC[C@]4(O)C(COC(CCC([O-])=O)=O)=O)=C1.[Na+]

InChI

1S/C25H34O8.Na/c1-23-9-7-15(26)11-14(23)3-4-16-17-8-10-25(32,24(17,2)12-18(27)22(16)23)19(28)13-33-21(31)6-5-20(29)30;/h11,16-18,22,27,32H,3-10,12-13H2,1-2H3,(H,29,30);/q;+1/p-1/t16-,17-,18-,22+,23-,24-,25-;/m0./s1

InChI key

HHZQLQREDATOBM-CODXZCKSSA-M

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Hydrocortisone for use in epithelial and endothelial adherent cell culture applications.

Application

Hydrocortisone 21-hemisuccinate sodium salt has been used as a supplement in Dulbecco′s modified Eagle medium/nutrient mixture F-12 (DMEM F12) for culturing oral squamous cell carcinoma (OSCC) cell lines and immortalized cone photoreceptor cell line.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manashi Priyadarshini et al.
Life sciences, 287, 120141-120141 (2021-11-15)
SARI (suppressor of activator protein (AP)-1, regulated by interferon (IFN) was identified as a novel tumor suppressor by applying subtraction hybridization to terminally differentiating human melanoma cells. The anti-tumor activity of SARI and the correlation between expression and cancer aggression
Ying Yuan et al.
Clinical epigenetics, 12(1), 135-135 (2020-09-08)
Hepatitis B virus covalently closed circular DNA (HBV cccDNA) is assembled by histones and non-histones into a chromatin-like cccDNA minichromosome in the nucleus. The cellular histone acetyltransferase GCN5, displaying succinyltransferase activity, is recruited onto cccDNA to modulate HBV transcription in
Kelsey A Hart et al.
American journal of veterinary research, 73(1), 68-75 (2011-12-30)
To compare daily endogenous cortisol production rate and the pharmacokinetics of an i.v. bolus of hydrocortisone between neonatal foals and adult horses. 10 healthy full-term 2- to 4-day-old foals and 7 healthy adult horses. Blood samples were collected from each
Crenguta Livia Calma et al.
Tropical doctor, 42(1), 54-55 (2011-11-08)
This case report focuses on hydatidosis, one of the most important parasitic zoonoses in Romania. We describe an interesting paediatric case of an eight-year old boy from a rural region presenting with a both giant and life-threatening pulmonary cyst (that
Naked & unconscious: crew's misdiagnosis could have cost patient her life.
Kimberly Doran
JEMS : a journal of emergency medical services, 37(10), 28-28 (2013-01-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service