Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hydrocortisone 21-hemisuccinate sodium salt
Synonym(s):
11β,17α,21-Trihydroxy-4-pregnene-3,20-dione 21-hemisuccinate sodium salt, Cortisol 21-hemisuccinate sodium salt
Empirical Formula (Hill Notation):
C
25
H
33
NaO
8
CAS Number:
125-04-2
Molecular Weight:
484.51
EC Number:
204-725-5
Products
Product Category
hormones (2)
bioactive small molecules (1)
Brand
Sigma-Aldrich (2)
Biological Source
Formula Weight
Gene Alias
Physical Form
Product Line
Purity
Research Area
Quality Segment
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
H2270
Hydrocortisone 21-hemisuccinate sodium salt,
powder, BioReagent, suitable for cell culture
Expand
View Pricing
H4881
Hydrocortisone 21-hemisuccinate sodium salt,
≥90% (HPLC)
Expand
View Pricing