W4128
Williams′ Medium E
With sodium bicarbonate, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture
Synonym(s):
Williams’ E medium
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.75
Recommended Products
Product Name
Williams′ Medium E, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Quality Level
sterility
sterile-filtered
form
liquid
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
components
L-glutamine: no
phenol red: 0.0107 g/L
glucose: 2.0 g/L (Dextro)
NaHCO3: 2.2 g/L
shipped in
ambient
storage temp.
2-8°C
General description
Williams′ Medium E has been shown to support the growth in long-term culture of adult liver epithelial cells.
Application
Williams′ Medium E has been used to culture primary human hepatocytes and to maintain hepatocellular carcinoma (JHH-6) cell lines.
Reconstitution
Supplement with 0.292 g/L L-glutamine.
also commonly purchased with this product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Targeted multicomponent polysomes for high efficiency, simultaneous anti-sense and gene delivery
Mandal S, et al.
Soft Matter, 7, 9424-9434 (2011)
Hannah L Paish et al.
Hepatology (Baltimore, Md.), 70(4), 1377-1391 (2019-04-10)
Precision cut liver slices (PCLSs) retain the structure and cellular composition of the native liver and represent an improved system to study liver fibrosis compared to two-dimensional mono- or co-cultures. The aim of this study was to develop a bioreactor
Florian Douam et al.
PLoS pathogens, 14(3), e1006908-e1006908 (2018-03-06)
Amino-acid coevolution can be referred to mutational compensatory patterns preserving the function of a protein. Viral envelope glycoproteins, which mediate entry of enveloped viruses into their host cells, are shaped by coevolution signals that confer to viruses the plasticity to
Isolation and long-term cell culture of epithelial-like cells from rat liver.
G M Williams et al.
Experimental cell research, 69(1), 106-112 (1971-11-01)
Charlie Colin-Pierre et al.
Biomolecules, 11(2) (2021-02-13)
The expression of glypicans in different hair follicle (HF) compartments and their potential roles during hair shaft growth are still poorly understood. Heparan sulfate proteoglycan (HSPG) distribution in HFs is classically investigated by conventional histology, biochemical analysis, and immunohistochemistry. In
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service