Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9031-34

Protein G High Performance Spintrap

Cytiva 28-9031-34, pack of 16 columns

Synonym(s):

Protein G Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.25

ligand

recombinant protein G lacking albumin-binding region

feature

wetted part: no

packaging

pack of 16 columns

manufacturer/tradename

Cytiva 28-9031-34

storage condition

(20% Ehtanol)
(20% Ethanol)

matrix

highly cross-linked 6% agarose

avg. part. size

34 μm

cleaning

2-9

working range

3-9

capacity

>1 mg binding capacity (lgG/column)(The binding capacity has been determined with human polyclonal IgG.)
>1 mg binding capacity(human IgG/column)

storage temp.

2-8°C

General description

pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.
pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.
protein G HP SpinTrap is a prepacked, single-use spin column containing protein G Sepharose High Performance.

Application

Protein G Sepharose HP has high affinity for the Fc region of IgG from a variety of species. The two options in the Protein enrichment protocol allow the attached antigen to be eluted separately (cross-link protocol) or together with the antibody (classic protocol).


There is also a protocol for efficient, small-scale purification of monoclonal and polyclonal antibodies from serum and cell culture supernatants in less than 20 min. No pretreatment of the sample is required.

Features and Benefits

  • For simple, small-scale purification of antibodies or enrichment of specific Proteins in prepacked spin columns that are ready to use.
  • Classic and cross-link protocols provide flexibility.
  • Elution conditions formatted for both electrophoresis and LC-MS analysis workflows.
  • Fast binding kinetics and high capacity provides high yield.
  • Prepacked with Protein G Sepharose High Performance for coupling of antibodies of IgG subclasses.
  • Easy scale-up with HiTrap® Protein G HP prepacked columns.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

HiTrap is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva
SpinTrap is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service